Thermo Scientific VSTAR00 Versa Star Meter Only
Representative image only.
Thermo Scientific Mfr # VSTAR00 – Item # EW-58825-80
Modularity allows you to configure the meter how you need it--for now and for later
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • 3 year warranty
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Buffer RecognitionDIN, NIST, and USA
  • Dissolved Oxygen Accuracy±0.2ppm from 0 to 20 ppm; ±0.5 ppm from 50 to 50 ppm
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Power (Hz)50 or 60
  • DescriptionVersa Star Meter Only
  • Warranty3 year
3 Year WarrantyIP54 Rated
Key Features
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • 3 year warranty
More About this Item
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
METER ONLY--Must purchase desired measuring modules and electrodes separately.