Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Choose from over 250 pages of products for Life Science research including electroporation products from BTX and Eppendorf. Request yours today.
Electroporation
Cole-Parmer offers a wide range of products for Life Science including products for electroporation. Our line of products include electroporation systems, cuvettes, buffers, and electrodes. Choose electroporation systems and products from such manufacturers as BTX®, Eppendorf®, and Thermo Scientific CelljecT. Our Application Specialists are available to help select the systems and products that suit your laboratory requirements.
Electroporation is a process by which a high voltage is applied across an electrical field to disrupt a cell membrane and introduce a foreign substance into the cell. This is done using a cuvette for electroporation and an electroporator. By introducing plasmids or foreign genes into the cell, alterations are made to the final cell, providing resistance to outside factors such as antibiotics. Once the process of electroporation is complete, a liquid medium is added to the reaction and cells are allowed to sit in an incubator for a period of time which allows the cells to recover and become resistant. The cells are then typically spread on agar plates for replication.
Electroporation Products
Life Science solutionsBrowse the NEW Life Science Products & Solutions Catalog (pdf)
  • Cell Biology & Microbiology
  • Lab Equipment & Supplies
    • Electroporators
    • Electroporation systems from BTX®, Thermo Scientific, and
      Eppendorf® including ElectroCell fusion and Multiporator®
    • Cuvettes for Electroporation
    • Sterile cuvettes in gap sizes of 1mm, 2mm, and 4mm for use in the
      transfection of bacteria, yeast, insect, plant, and mammalian cells
    • Electroporation Buffers
    • Hypo-osmolar and Iso-osmolar buffers for
      transfection of mammalian cells
    • Microslides and Chamber
    • Microslides in gap sizes ranging from 0.5mm to 10mm
      and meander fusion chambers for electroporators
    • Specialty Electrodes
    • Caliper electrodes and petri dish electrodes for
      use with electroporation systems
    • Prepared Media Products
    • Prepared media including lactose broth, potato dextrose agar,
      and TSA in bottles, tubes, and plate formats
    • Pipettes and Tips
    • Multichannel and single channel pipettes with adjustable or set
      volume and a wide assortment of tips for all requirements
    • Incubators
    • Benchtop, CO2, general-purpose, hybridization, shaking,
      and refrigerated incubators in a variety of sizes and specifications
    Cole-Parmer
    625 East Bunker Court
    Vernon Hills, Illinois 60061
    Phone: 1-800-323-4340
    Fax: 1-847-247-2929
    E-Mail: info@Coleparmer.com
    Copyright © 2010
    Cole-Parmer Sites  Feedback
    Cole-Parmer:
    United States
    Canada
    China
    United Kingdom
    India
    McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams