Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
New Oakton® Waterproof Handheld
600 Series Meters
New Oakton® waterproof handheld 600 series meters
Expanded range, resolution, and accuracy, plus increased memory and advanced communications!
  • No more excuses for missing data! Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
These waterproof handheld meters feature a -2.00 to 19.99 pH range expandable to 0.001 pH resolution. Combined with accuracies up to 0.002 pH, these meters provide a new benchmark in our pH meter line. Store up to 500 data sets in the meter’s memory. Time-and-date-stamping meets Good Laboratory Practice (GLP) standards. The pH 600 meters offer the latest in calibration capabilities including up to six calibration points, a user-settable calibration due alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards. The infrared IrDA wireless technology makes PC downloading more convenient and easier. Dual power up allows you to use batteries while out in the field or the optional AC power adapter while in the lab. As always, our waterproof meters are IP67-rated for both dust and water intrusion. Even with no probes attached!
The new Oakton® Waterproof Handheld 600 Series Meters will be available in early May.
Get your free copy of our 2006 Water Quality Catalog with complete specifications and ordering information.
Oakton® Waterproof pH 600 Meter
Oakton® Waterproof pH 610 Meter
Oakton® Waterproof pH/Ion 620 Meter
Complete
Product Index
Oakton® Waterproof pH 600 Meter
Oakton® Waterproof pH 610 Meter
Oakton® Waterproof pH/Ion 620 Meter
More Oakton® pH Meters