Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
       
       
Home Products & Services Technical Library myCP Account Contact/About Us

Fluid handling solutions from Masterflex


Tubing Pumps
  • C/L® (Compact/LowFlow)
  • L/S® (Lab Standard)
  • I/P® (Industrial Process)
  • B/T® (Batch Transfer)

    Masterflex Tubing

    Liqui-Sense® Emergency
    Leak Detection System




  • Request your FREE Masterflex Encyclopedia Vol3!


    Need help selecting system components that work together? Try this...
    Masterflex Technical Info
  • Manuals
  • Tubing Performance Data
  • Pumphead Torque
        Requirements
  • Vacuum/Suction Lift Data
  • Dimensional Drawings
    Go To
    Masterflex.com/techinfo
  • Cole-Parmer general catalogue request Filling all your fluid process needs...

    Request the latest
    Cole-Parmer General Catalog today!


    Tech Tools___________________________________________
    Find it FAST!
    Choose a Parametric Search:
    Chemical Compatibility Database
  • General
  • Masterflex Tubing &     Chemical
  • Masterflex Tubing & Food
  • Masterflex Rollers

  • Masterflex Tubing Pumps
    Application Solutions
    Fluid Handling Technical Articles
    Application Assistance
    Need help from our experts, use this form.

    Standard Pipe Thread Fittings

    Technical Equations

    Viscosity Values