Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
       
       
Home Products & Services Technical Library myCP Account Contact/About Us




Tubing Pumps
  • C/L® (Compact/LowFlow)
  • L/S® (Lab Standard)
  • I/P® (Industrial Process)
  • B/T® (Batch Transfer)

    Masterflex Tubing

    Liqui-Sense® Emergency
    Leak Detection System




  • Fluid Handling Products

    Aseptic Sampling

    Filtration

    Fittings

    Flowmeters

    Hose and Hose Fittings

    Ismatec Peristaltic Pumps

       Ismatec tubing

    Level Control

    Microfluidics

    Mixers (Overhead stirrers)

    Piping

    Pressure Gauges

    Pumps
      Centrifugal pumps
      Drum pumps
      Filler pumps
      Gear pumps
      Industrial hose pumps
      Positive displacement pumps
      Vacuum pumps

    Sanitary

    Tanks

    Tubing

    Valves


    Cole-Parmer general catalogue request Filling all your fluid process needs...

    Request the latest
    Cole-Parmer General Catalog today!
    Tech Tools___________________________________________
    Find it FAST!
    Choose a Parametric Search:
    Chemical Compatibility Database
  • General
  • Masterflex Tubing &     Chemical
  • Masterflex Tubing & Food
  • Masterflex Rollers
  • Masterflex Tubing Pumps
    Application Solutions
    Fluid Handling Technical Articles
    Application Assistance
    Need help from our experts, use this form.

    Standard Pipe Thread Fittings

    Technical Equations

    Viscosity Values

    Need help selecting system components that work together? Try this...
    Masterflex Technical Info
  • Manuals
  • Tubing Performance Data
  • Pumphead Torque
        Requirements
  • Vacuum/Suction Lift Data
  • Dimensional Drawings
    Go To
    Masterflex.com/techinfo
    NIST-traceable calibration and certificate, air/gas flowmeters with test data

    NIST-traceable calibration and certificate, mass flowmeters with test data