This product is now discontinued, View Related Products
Adapt-A-Port Kits 4103
Standard taper is threaded to accept two interchangeable ports—one with hose barb for vacuum tubing; one for thermometers, glass tubing, and pipettes. Ports are 6 mm. Viton® O-rings ensure a tight seal.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Taper Size
Availability Pricing
Scienceware 20304-1560 Adapt-a-port kit, size 24/40 EW-06139-55 Mfr # 20304-1560
COMPARE
24/40
Call For Price
Scienceware 20304-1570 adapt-a-port kit, size 34/45 EW-06139-65 Mfr # 20304-1570
COMPARE
34/45