Material Safety Data Sheet
Diethyl Sulfite, 98%
ACC# 78733
Section 1 - Chemical Product and Company Identification |
MSDS Name: Diethyl Sulfite, 98%
Catalog Numbers:
AC209930000, AC209930100
Synonyms:
Diethyl sulphite; Ethyl sulfite; Sulfurous acid, diethyl ester
Company Identification:
Acros Organics N.V.
One Reagent Lane
Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300
Section 2 - Composition, Information on Ingredients
|
CAS# |
Chemical Name |
Percent |
EINECS/ELINCS |
623-81-4 |
Diethyl Sulfite |
98 | 210-815-5 |
Section 3 - Hazards Identification
|
EMERGENCY OVERVIEW
Appearance: clear, colorless liquid. Flash Point: 53 deg C.
Warning! Flammable liquid and
vapor. Causes respiratory tract irritation. Irritant.
May cause digestive tract irritation. May cause central nervous
system depression.
Target Organs: Central nervous system.
Potential Health Effects
Eye:
Causes eye irritation. May cause chemical conjunctivitis and
corneal damage.
Skin:
Causes skin irritation. May cause dermatitis. May cause cyanosis of
the extremities.
Ingestion:
Causes gastrointestinal irritation with nausea, vomiting and
diarrhea. Ingestion of large amounts may cause CNS depression.
Inhalation:
Causes respiratory tract irritation. Vapors may cause dizziness or
suffocation. Can produce delayed pulmonary edema. May cause burning
sensation in the chest.
Chronic:
Effects may be delayed.
Section 4 - First Aid Measures
|
Eyes:
Immediately flush eyes with plenty of water for at least 15 minutes,
occasionally lifting the upper and lower eyelids. Get medical aid.
Skin:
Get medical aid. Flush skin with plenty of water for at least 15
minutes while removing contaminated clothing and shoes. Wash clothing
before reuse.
Ingestion:
Never give anything by mouth to an unconscious person. Get medical
aid. Do NOT induce vomiting. If conscious and alert, rinse mouth and
drink 2-4 cupfuls of milk or water.
Inhalation:
Remove from exposure and move to fresh air immediately. If not
breathing, give artificial respiration. If breathing is difficult,
give oxygen. Get medical aid.
Notes to Physician:
Treat symptomatically and supportively.
Section 5 - Fire Fighting Measures
|
General Information:
As in any fire, wear a self-contained breathing apparatus in
pressure-demand, MSHA/NIOSH (approved or equivalent), and full
protective gear. Vapors may form an explosive mixture with air.
Vapors can travel to a source of ignition and flash back. During a
fire, irritating and highly toxic gases may be generated by thermal
decomposition or combustion. Will burn if involved in a fire. Use
water spray to keep fire-exposed containers cool. Containers may
explode in the heat of a fire. Flammable liquid and vapor. Vapors
may be heavier than air. They can spread along the ground and
collect in low or confined areas.
Extinguishing Media:
For small fires, use dry chemical, carbon dioxide, water spray or
alcohol-resistant foam. For large fires, use water spray, fog, or
alcohol-resistant foam. Use water spray to cool fire-exposed
containers. Water may be ineffective. Do NOT use straight streams of
water.
Flash Point: 53 deg C ( 127.40 deg F)
Autoignition Temperature: Not available.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating:
(estimated) Health: 1; Flammability: 2; Instability: 0
Section 6 - Accidental Release Measures
|
General Information: Use proper personal protective equipment as indicated
in Section 8.
Spills/Leaks:
Clean up spills immediately, observing precautions in the Protective
Equipment section. Remove all sources of ignition. Absorb spill using
an absorbent, non-combustible material such as earth, sand, or
vermiculite. Do not use combustible materials such as sawdust. Use a
spark-proof tool. Provide ventilation. A vapor suppressing foam may
be used to reduce vapors.
Section 7 - Handling and Storage
|
Handling:
Ground and bond containers when transferring material. Use
spark-proof tools and explosion proof equipment. Avoid contact with
eyes, skin, and clothing. Empty containers retain product residue,
(liquid and/or vapor), and can be dangerous. Keep container tightly
closed. Keep away from heat, sparks and flame. Avoid ingestion and
inhalation. Use with adequate ventilation. Wash clothing before
reuse. Do not pressurize, cut, weld, braze, solder, drill, grind, or
expose empty containers to heat, sparks or open flames.
Storage:
Keep away from sources of ignition. Store in a tightly closed
container. Store in a cool, dry, well-ventilated area away from
incompatible substances. Flammables-area.
Section 8 - Exposure Controls, Personal Protection
|
Engineering Controls:
Facilities storing or utilizing this material should be equipped
with an eyewash facility and a safety shower. Use adequate general or
local explosion-proof ventilation to keep airborne levels to
acceptable levels.
Exposure Limits
Chemical Name |
ACGIH |
NIOSH |
OSHA - Final PELs |
Diethyl Sulfite
|
none listed
|
none listed
|
none listed
|
OSHA Vacated PELs:
Diethyl Sulfite:
No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes:
Wear appropriate protective eyeglasses or chemical
safety goggles as described by OSHA's eye and face
protection regulations in 29 CFR 1910.133 or European
Standard EN166.
Skin:
Wear appropriate protective gloves to prevent skin
exposure.
Clothing:
Wear appropriate protective clothing to prevent skin
exposure.
Respirators:
A respiratory protection program that meets OSHA's 29
CFR 1910.134 and ANSI Z88.2 requirements or European
Standard EN 149 must be followed whenever workplace
conditions warrant respirator use.
Section 9 - Physical and Chemical Properties
|
Physical State: Liquid
Appearance: clear, colorless
Odor: None reported.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: 4.8
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: 158 - 160 deg C @ 760.00mmHg
Freezing/Melting Point:Not available.
Decomposition Temperature:Not available.
Solubility: Not available.
Specific Gravity/Density:1.8830g/cm3
Molecular Formula:C4H10O3S
Molecular Weight:138.18
Section 10 - Stability and Reactivity
|
Chemical Stability:
Stable at room temperature in closed containers under normal storage
and handling conditions.
Conditions to Avoid:
Incompatible materials, ignition sources, excess heat, strong
oxidants.
Incompatibilities with Other Materials:
Strong oxidizing agents.
Hazardous Decomposition Products:
Carbon monoxide, irritating and toxic fumes and gases, carbon
dioxide.
Hazardous Polymerization: Has not been reported.
Section 11 - Toxicological Information
|
RTECS#:
CAS# 623-81-4:
WT3511000
LD50/LC50:
Not available.
Carcinogenicity:
CAS# 623-81-4:
Not listed by ACGIH, IARC, NTP, or CA Prop 65.
Epidemiology:
No data available.
Teratogenicity:
No data available.
Reproductive Effects:
No data available.
Mutagenicity:
No data available.
Neurotoxicity:
No data available.
Other Studies:
Section 12 - Ecological Information
|
No information available.
Section 13 - Disposal Considerations
|
Chemical waste generators must determine whether a discarded chemical is classified
as a hazardous waste.
US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3.
Additionally, waste generators must consult state and local hazardous waste regulations to
ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.
Section 14 - Transport Information
|
|
US DOT |
Canada TDG |
Shipping Name: |
DOT regulated - small quantity provisions apply (see 49CFR173.4) |
No information available. |
Hazard Class: |
|
|
UN Number: |
|
|
Packing Group: |
|
|
Section 15 - Regulatory Information
|
US FEDERAL
TSCA
CAS# 623-81-4 is listed on the TSCA inventory.
Health & Safety Reporting List
None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
None of the chemicals in this product have a TPQ.
Section 313
No chemicals are reportable under Section 313.
Clean Air Act:
This material does not contain any hazardous air pollutants.
This material does not contain any Class 1 Ozone depletors.
This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
None of the chemicals in this product
are listed as Hazardous Substances under the CWA.
None of the chemicals in this
product are listed as Priority Pollutants under the CWA.
None of the chemicals in this
product are listed as Toxic Pollutants under the CWA.
OSHA:
None of the chemicals in this product
are considered highly hazardous by OSHA.
STATE
CAS# 623-81-4 is not present on
state lists from CA, PA, MN, MA, FL, or NJ.
California Prop 65
California No Significant Risk Level:
None of the chemicals in this product are listed.
European/International Regulations
European Labeling in Accordance with EC Directives
Hazard Symbols:
XI
Risk Phrases:
R 10 Flammable.
R 36/37/38 Irritating to eyes, respiratory system and skin.
Safety Phrases:
S 16 Keep away from sources of ignition - No smoking.
S 24/25 Avoid contact with skin and eyes.
S 33 Take precautionary measures against static discharges.
S 9 Keep container in a well-ventilated place.
WGK (Water Danger/Protection)
CAS# 623-81-4: No information available.
Canada - DSL/NDSL
CAS# 623-81-4 is listed on Canada's NDSL List.
Canada - WHMIS
This product has a WHMIS classification of B2, D2B.
This product has been classified in accordance with the hazard
criteria of the Controlled Products Regulations and the MSDS
contains all of the information required by those regulations.
Canadian Ingredient Disclosure List
Section 16 - Additional Information
|
MSDS Creation Date: 12/22/1998
Revision #6 Date: 10/03/2005
The information above is believed to be accurate and represents the best
information currently available to us. However, we make no warranty of
merchantability or any other warranty, express or implied, with respect to
such information, and we assume no liability resulting from its use. Users
should make their own investigations to determine the suitability of the
information for their particular purposes. In no event shall Fisher be liable
for any claims, losses, or damages of any third party or for lost profits
or any special, indirect, incidental, consequential or exemplary
damages, howsoever arising, even if Fisher has been advised of
the possibility of such damages.