|
|
|
|
| CAS# | Chemical Name | Percent | EINECS/ELINCS |
| 7783-85-9 | Ammonium Iron (II) Sulfate Hexahydrate | 100.0 | unlisted |
|
|
Appearance: light green powder.
Warning! Causes respiratory tract irritation.
Irritant. Causes eye and skin irritation. May cause digestive tract
irritation. Light sensitive. Air sensitive. May cause liver damage.
May cause cardiac disturbances.
Target Organs: Liver, cardiovascular system.
Potential Health Effects
Eye:
Causes eye irritation. May cause chemical conjunctivitis.
Skin:
Causes skin irritation.
Ingestion:
May cause gastrointestinal irritation with nausea, vomiting and
diarrhea. May cause liver damage. May cause cardiac disturbances.
Inhalation:
Causes respiratory tract irritation. Can produce delayed pulmonary
edema.
Chronic:
Effects may be delayed. Chronic exposure may cause liver damage.
|
|
|
|
|
|
|
|
|
|
| Chemical Name | ACGIH | NIOSH | OSHA - Final PELs |
| Ammonium Iron (II) Sulfate Hexahydrate | 1 mg/m3 TWA (as Fe) (listed under Iron salts (soluble)). | 1 mg/m3 TWA (as Fe) (listed under Iron salts (soluble)). | none listed |
|
|
|
|
|
|
|
|
|
|
|
|
| US DOT | Canada TDG | |
| Shipping Name: | Not regulated as a hazardous material | No information available. |
| Hazard Class: | ||
| UN Number: | ||
| Packing Group: |
|
|
|
|