Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Product Type:
Availability:
Form:
Liquid (164)
Crystal (36)
Powder (20)
Packaging:
Quantity (g):
500 (47)
2500 (22)
100 (3)
125 (3)
12000 (3)
25 (1)
50000 (1)
3000 (1)
Quantity (mL):
4000 (41)
1000 (34)
500 (33)
3800 (19)
20000 (16)
2500 (10)
950 (3)
473 (3)
19000 (2)
2200 (2)
100 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

 
 Product Detail
Sort
 Availability
 Price
chemicals_generic.jpg - Potassium Iodide Granular Acs 500g

Potassium Iodide Granular Acs 500g

Product Type Chemical Reagents,Purity grade ACS,Form Crystal
EW-89100-47
Usually ships in 6 days.
$205.00 USD/EACH

Qty:

chemicals_generic.jpg - Hydrogen Peroxide 30% Acs 4L

Hydrogen Peroxide 30% Acs 4L

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-88
Usually ships in 16 days.
$286.00 USD/EACH

Qty:

chemicals_generic.jpg - ETHANOL, ANHYDROUS DENATURED 200 Proof   4L

ETHANOL, ANHYDROUS DENATURED 200 Proof 4L

Product Type Chemical Reagents,Form Liquid,Packaging Poly Bottle
EW-89000-59
Usually ships in 6 days.
$82.50 USD/EACH

Qty:

chemicals_generic.jpg - Reagent Alcohol Absolute 4L poly

Reagent Alcohol Absolute 4L poly

Product Type Chemical Reagents,Form Liquid,Packaging Poly Bottle
EW-89100-58
Usually ships in 18 days.
$63.75 USD/EACH

Qty:

chemicals_generic.jpg - Isopropyl Alcohol Hplc 4L

Isopropyl Alcohol Hplc 4L

Product Type Chemical Reagents,Purity grade HPLC,Form Liquid
EW-89000-96
Usually ships in 8 days.
$105.00 USD/EACH

Qty:
See all 2-Propanol

chemicals_generic.jpg - ETHANOL, ANHYDROUS DENATURED 200 Proof  4L

ETHANOL, ANHYDROUS DENATURED 200 Proof 4L

Product Type Chemical Reagents,Form Liquid,Packaging Poly Bottle
EW-89000-60
Usually ships in 9 days.
$75.00 USD/EACH

Qty:

chemicals_generic.jpg - ETHANOL, ANHYDROUS DENATURED 200 Proof   1 Gallon

ETHANOL, ANHYDROUS DENATURED 200 Proof 1 Gallon

Product Type Chemical Reagents,Form Liquid,Packaging Poly Bottle
EW-89000-58
Usually ships in 6 days.
$77.00 USD/EACH

Qty:

AGROS22214.jpg - Acetic Acid Glacial Acs 2.5l gls

Acetic Acid Glacial Acs 2.5l gls

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-02
Usually ships in 7 days.
$57.00 USD/EACH

Qty:
See all Acetic Acid

chemicals_generic.jpg - Methanol, Anhydrous Acs 4L  acs poly gls

Methanol, Anhydrous Acs 4L acs poly gls

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89100-08
Usually ships in 8 days.
$66.00 USD/EACH

Qty:
See all Methanol

chemicals_generic.jpg - Isopropyl Alcohol Acs 4L

Isopropyl Alcohol Acs 4L

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-92
Usually ships in 7 days.
$85.00 USD/EACH

Qty:
See all Isopropanol

chemicals_generic.jpg - Isopropyl Alcohol Acs 20l

Isopropyl Alcohol Acs 20l

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-93
Usually ships in 10 days.
$285.00 USD/EACH

Qty:
See all Isopropanol

AGROS41935.jpg - Silver Nitrate Acs 125g

Silver Nitrate Acs 125g

Product Type Chemical Reagents,Purity grade ACS,Form Crystal
EW-89100-61
Usually ships in 8 days.
$350.00 USD/EACH

Qty:
See all Silver Nitrate

AGROS41935.jpg - Silver Nitrate Acs 500g

Silver Nitrate Acs 500g

Product Type Chemical Reagents,Purity grade ACS,Form Crystal
EW-89100-60
Usually ships in 10 days.
$975.00 USD/EACH

Qty:
See all Silver Nitrate

chemicals_generic.jpg - Sodium Hydroxide 0.10n   4L

Sodium Hydroxide 0.10n 4L

Product Type Chemical Reagents,Form Liquid,Packaging Poly Bottle
EW-89200-62
Usually ships in 8 days.
$36.50 USD/EACH

Qty:

chemicals_generic.jpg - Nitric Acid 70% Acs 2.2L

Nitric Acid 70% Acs 2.2L

Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89100-18
Usually ships in 7 days.
$71.00 USD/EACH

Qty:
See all Nitric Acid

245 results (showing 1 - 15 )
Per Page 
View products by: Job Title | Category | Brand | Part Number | Price
View product categories by: Popularity | Favorites | Job Title | Price | Part Number
View popular items by: Popularity | Our Choices | All-Around Favorites | Job Title
View reviews by: Popularity | Product Title | Best Rated