United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Shop by Industry
(2389)
Lab Supplies & Consumables
(1206)
Tubing, Fittings, and Hose
(1168)
[+]
Show more
(23)
Laboratory Equipment
(460)
Fluid & Liquid Handling
(357)
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
(338)
Life Science
(215)
Flow, Level & Valves
(185)
General Accessories
(140)
Pumps
(137)
Masterflex® Peristaltic Pumps
(106)
Chemicals, Reagents, & Lubricants
(106)
Temperature, Humidity and Moisture Products
(73)
Pressure & Vacuum Instruments
(62)
Balances & Scales
(47)
Safety Products
(37)
Process Equipment & Instrumentation
(24)
Microscopes & Magnification
(24)
Heating Equipment
(12)
Cleaning & Janitorial Supplies
(11)
Furniture & Fume Hoods
(7)
Calibration
(6)
Recorders & Data Acquisition
(5)
Clean Room
(2)
Material Testing & Physical Testing
(1)
Material Storage & Handling
(1)
[-]
Show less
Brand:
Cole-Parmer
(538)
Masterflex
(410)
Oakton
(165)
[+]
Show more
(17)
Colder
(111)
Thermo Scientific
(71)
Tygon
(69)
C-FLEX
(51)
Ashcroft
(39)
Corning
(33)
Thermo Scientific Orion
(32)
Mono-Mold
(21)
B-D
(21)
Thermo Scientific Nalgene
(20)
Pyrex
(19)
StableTemp
(17)
Myron L
(17)
Viton
(13)
Ohaus
(11)
Eutech Instruments
(10)
Glove Bag
(6)
[-]
Show less
Product Type:
Pump Tubing
(340)
Luer fittings
(180)
Accessory
(152)
[+]
Show more
(17)
Barbed fittings
(147)
Tubing
(118)
Quick Disconnect fittings
(112)
Tubing (Silicone)
(71)
Beaker
(61)
Bottles
(57)
Pump Head
(56)
Compression fittings
(55)
Filter paper
(46)
Testing Sieves
(43)
Calibration Solutions
(40)
Carboys / Jerricans / Jugs
(40)
Ball Valves (manual)
(36)
pH Electrodes
(33)
pH Meters
(28)
Weighing dishes
(23)
Toploading Balance
(22)
[-]
Show less
Price Range:
$0 to $100
(2110)
$100 to $299
(775)
$300 to $499
(155)
[+]
Show more
(4)
$500 to $799
(114)
$800 to $1,099
(46)
$1,100 to $1,999
(35)
$2,000 and Over
(21)
[-]
Show less
Availability:
In Stock
(3236)
Learn more:
Isolating Genomic DNA from Whole Blood
Basics of Cartridge Filtration
Equipping a Laboratory: A Study in Customer Service Excellence
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
[+]
Show More
ISO 80369It’s Coming to a Device Maker Near You
Bitter Brew
Physical Properties of Glass
Fittings Selection Guide
Electron zap turns flimsy plastic into sturdy shrink wrap
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Physical Properties of Stainless Steel
Tubing Selection Guide
ChemicalsSpecial Ordering Information
ChemicalsSpecial Ordering Information
Decrease bacterial growth in your tubin
Cole-Parmers Bioconnect Program
Physical Properties of Plastics
Mixer Selection Guide
Ismatec: Pump Selection Guide
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Shop by
>
Brand - Guaranteed In Stock Program
Guaranteed In Stock Program
We understand the importance of having products available when you need them. Listed below are the specific items we guarantee to be In Stock or we will pay the shipping costs- guaranteed! GIS items listed are guaranteed in stock through 12/31/11.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Digital Drive, 600 RPM, 115/230 VAC
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.006
EW-07523-80
In stock.
$1,830.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex L/S Standard Digital Drives
Masterflex L/S Digital Drive, 100 RPM, 115/230 VAC
Product Type Pump Drive,Max flow rate mL/min 566,Min flow rate mL/min 0.001
EW-07523-90
In stock.
$1,830.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex L/S Standard Digital Drives
Masterflex
platinum-cured silicone tubing, L/S
15, 25 ft.
Product Type Pump Tubing,Pump Series L/S,Tubing formulation Silicone
EW-96410-15
In stock.
$81.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High-Performance Precision Silicone (platinum cured) Tubing
Masterflex
platinum-cured silicone tubing, I/P
73, 25 ft.
Product Type Pump Tubing,Pump Series I/P,Tubing formulation Silicone
EW-96410-73
In stock.
$161.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Precision Silicone (platinum cured) Tubing
Masterflex
platinum-cured silicone tubing, L/S
24, 25 ft.
Product Type Pump Tubing,Pump Series L/S,Tubing formulation Silicone
EW-96410-24
In stock.
$103.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High-Performance Precision Silicone (platinum cured) Tubing
Masterflex
I/P
Precision Brushless Drive, 33 to 650 rpm, 115/230 VAC
Product Type Masterflex I/P Drive,Max flow rate mL/min 19000,Min flow rate mL/min 200
EW-77410-10
In stock.
$1,960.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P
Precision Brushless Drives
Masterflex
platinum-cured silicone tubing, L/S
36, 25 ft.
Pump Series L/S,Product Type Pump Tubing,Tubing formulation Silicone
EW-96410-36
In stock.
$143.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High-Performance Precision Silicone (platinum cured) Tubing
Masterflex
L/S
economy variable-speed drive, 20 to 600 rpm, 115 VAC.
Product Type Pump Drive,Max flow rate mL/min 2900,Min flow rate mL/min 1.2
EW-07554-90
In stock.
$610.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Economy Variable-Speed Console Drives
Masterflex
L/S
Easy-Load
II pump head for high-performance precision tubing, fixed occlusion, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77200-62
In stock.
$285.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Easy-Load
II Pump Heads for High-Performance Precision Tubing
Masterflex
L/S
Easy-Load
pump head for precision tubing; PSF housing, CRS rotor.
Product Type Pump Head,Rotor material Cold-rolled steel,Housing material Polysulfone
EW-07518-00
In stock.
$198.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Easy-Load Pump Heads for Precision Tubing
Masterflex
I/P
Easy-Load
pump head, PSF housing/SS rotor.
Product Type Pump Head,Max flow rate LPM 13.0,Max flow rate GPM 3.5
EW-77601-10
In stock.
$636.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P
Easy-Load
Pump Heads
Masterflex
L/S
Digital Pump System with Easy-Load
II Pump Head, 600 RPM, 115/230 VAC
Product Type Peristaltic Pump,Pump Series L/S,Min flow rate mL/min 0.28
EW-77921-70
In stock.
$2,145.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Standard Digital Pump Systems
Masterflex
L/S
Easy-Load
II pump head for precision tubing, fixed occlusion, SS rotor.
Product Type Pump Head,Rotor material Stainless steel,Housing material Polyphenylene sulfide
EW-77200-60
In stock.
$285.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Easy-Load
II Pump Heads for Precision Tubing
Oakton
Waterproof EcoTestr
pH 2 Pocket pH Tester
Product Type pH Meters,Meter style Portable,Dimensions " W 1-5/8
EW-35423-10
In stock.
$61.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Waterproof EcoTestr
pH 2 Pocket pH Tester
Oakton Waterproof pHTestr 30 Pocket pH Tester
Meter style Tester,Temperature compensation Automatic,CE Compliance Yes
EW-35634-30
Usually ships in 44 days or sooner.
$115.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Waterproof pHTestr
30 Pocket pH Tester
3242
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated