Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Search within results:
Go
Category:
Fittings (793)
Brand:
Cole-Parmer (1025)
Product Type:
Availability:
In Stock (888)
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 
Home > Cole-Parmer Catalog > search results for 'Ammonium Chloride Buffer Solution pH 10 0 EP Buffer Solution 4007300'

Showing 5454 results for 'Ammonium Chloride Buffer Solution pH 10 0 EP Buffer Solution 4007300'

Products
Technical Resources
 Product Detail
Sort
 Availability
 Price

Ammonium Chloride Buffer Solution, pH 10.0, EP Buffer Solution 4007300 (500 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 12125-02-9, 1336-21-6 and 7732-18-5
EW-86969-65
Available for order
or call 1-888-358-4705 to order
$58.50 USD/EACH

Qty:

Oakton pH Calibration Buffer, 10.00, 500 mL

  • Product Type: pH Buffers
  • Volume: 500 mL
  • Description: pH 10.00
EW-00654-08
In stock.
$13.00 USD/PINT

Qty:

Oakton pH Calibration Buffer, 10.00, 1 Gal

  • Product Type: pH Buffers
  • Volume: 4000 mL
  • Description: pH 10.00
EW-05942-64
In stock.
$44.25 USD/EACH

Qty:

Oakton pH Calibration Buffer, 10.00, 1.0 L

  • Product Type: pH Buffers
  • Volume: 1000 mL
  • Description: pH 10.00
EW-05942-62
In stock.
$22.25 USD/EACH

Qty:

Oakton 10.00 pH buffer, 500 mL, case of 12

  • Product Type: pH Buffers
  • Volume: 500 mL
  • Description: pH 10.00
EW-05942-61
In stock.
$117.00 USD/CASE of 12

Qty:

Oakton pH Calibration Buffer, 10.00, 1 Gal, case of 4

  • Product Type: pH Buffers
  • Volume: 4000 mL
  • Description: pH 10.00
EW-05942-65
In stock.
$161.00 USD/CASE of 4

Qty:

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (1 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7732-18-5, 497-19-8, 144-55-8, 1310-73-2
  • Form: Liquid
EW-88400-21
Available for order
or call 1-888-358-4705 to order
$33.50 USD/EACH

Qty:

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, 1 gallon bottle

  • Product Type: Inorganic Chemicals
  • CAS Number: 7732-18-5, 497-19-8, 144-55-8, 1310-73-2
  • Form: Liquid
EW-88400-19
Available for order
or call 1-888-358-4705 to order
$72.25 USD/EACH

Qty:

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (1 gal)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7732-18-5, 7558-79-4, 7778-77-0
  • Form: Liquid
EW-88400-13
Available for order
or call 1-888-358-4705 to order
$66.75 USD/EACH

Qty:

Buffer, pH 7.00 ± 0.01 at 25C (Color Coded Yellow), 4L poly bottle

  • Product Type: Inorganic Chemicals
  • CAS Number: 7732-18-5, 7558-79-4, 7778-77-0
  • Form: Liquid
EW-88400-15
Usually ships in 12 days.
$65.50 USD/EACH

Qty:

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (500 mL)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7732-18-5, 7558-79-4, 7778-77-0
  • Form: Liquid
EW-88400-14
Available for order
or call 1-888-358-4705 to order
$19.25 USD/EACH

Qty:

Buffer Solution, pH 8.00 (Certified) (1 liter)

  • Product Type: Inorganic Chemicals
  • Purity grade: Certified
  • CAS Number: 7778-77-0 and 1310-73-2 and 7732-18-5
EW-88084-17
Usually ships in 6 days.
$82.25 USD/EACH
$47.94 USD/EACH

Qty:

Buffer Solution, pH 6.00 (Certified) (1 liter)

  • Product Type: Inorganic Chemicals
  • Purity grade: Certified
  • CAS Number: 7778-77-0 and 1310-73-2 and 7732-18-5
EW-88084-03
Usually ships in 3 days.
$97.00 USD/EACH
$57.00 USD/EACH

Qty:

Buffer Solution, pH 3.5 R (32 oz) (Chemical Buffers)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 631-61-8, 7647-01-0 and 7732-18-5
EW-86978-46
Available for order
or call 1-888-358-4705 to order
$90.75 USD/EACH

Qty:

Buffer Solution, pH 3.5 R (500 mL) (Chemical Buffers)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 631-61-8, 7647-01-0 and 7732-18-5
EW-86970-39
Available for order
or call 1-888-358-4705 to order
$54.50 USD/EACH

Qty:

5454 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#