Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Search within results:
Go
Category:
Brand:
Cole-Parmer (2615)
Product Type:
Availability:
In Stock (2823)
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 
Home > Cole-Parmer Catalog > search results for 'Hydrochloric Acid 50 v v Aqueous Solution 1 1'

Showing 13691 results for 'Hydrochloric Acid 50 v v Aqueous Solution 1 1'

Products
Technical Resources
 Product Detail
Sort
 Availability
 Price

Hydrochloric Acid, 50% (v/v) Aqueous Solution (1 + 1) (4 L)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86986-03
Usually ships in 5 days.
$110.00 USD/EACH

Qty:

Hydrochloric Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86972-72
Available for order
or call 1-888-358-4705 to order
$41.25 USD/EACH

Qty:

Hydrochloric Acid, 50% (v/v) Aqueous Solution (1 + 1) (32 oz)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86980-01
Available for order
or call 1-888-358-4705 to order
$47.50 USD/EACH

Qty:

Hydrochloric Acid, 1.96% (v/v) Aqueous Solution (1 + 50) (1 L)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86979-94
Usually ships in 10 days.
$24.00 USD/EACH

Qty:

Hydrochloric Acid, 1.96% (v/v) Aqueous Solution (1 + 50) (500 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86972-65
Usually ships in 10 days.
$16.00 USD/EACH

Qty:

Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (2.5 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 7664-93-9 and 7732-18-5
  • Form: Liquid
EW-86984-46
Usually ships in 14 days.
$111.00 USD/EACH

Qty:

Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 7664-93-9 and 7732-18-5
  • Form: Liquid
EW-86976-69
Available for order
or call 1-888-358-4705 to order
$31.25 USD/EACH

Qty:

Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (20 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 7664-93-9 and 7732-18-5
  • Form: Liquid
EW-86994-53
Available for order
or call 1-888-358-4705 to order
$309.00 USD/EACH

Qty:

Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (1 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 7664-93-9 and 7732-18-5
  • Form: Liquid
EW-86983-55
Usually ships in 5 days.
$46.25 USD/EACH

Qty:

Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (1 gal)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 7664-93-9 and 7732-18-5
  • Form: Liquid
EW-86988-23
Usually ships in 7 days.
$126.00 USD/EACH

Qty:

Acetic Acid, 50% (v/v) Aqueous Solution (1 + 1) (4 L)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 64-19-7 and 7732-18-5
EW-86984-60
Available for order
or call 1-888-358-4705 to order
$110.00 USD/EACH

Qty:

Acetic Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 64-19-7 and 7732-18-5
EW-86969-17
Available for order
or call 1-888-358-4705 to order
$44.75 USD/EACH

Qty:

Acetic Acid, 50% (v/v) Aqueous Solution (1 + 1) (32 oz)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 64-19-7 and 7732-18-5
EW-86977-61
Usually ships in 8 days.
$67.25 USD/EACH

Qty:

Hydrochloric Acid, 10% (v/v) Aqueous Solution (1 + 9) (1 gal)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86986-00
Available for order
or call 1-888-358-4705 to order
$57.75 USD/EACH

Qty:

Hydrochloric Acid, 10% (v/v) Aqueous Solution (1 + 9) (1 L)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7647-01-0 and 7732-18-5
EW-86979-98
Usually ships in 6 days.
$32.25 USD/EACH

Qty:

13691 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#