Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Search within results:
Go
Category:
Fittings (1140)
Brand:
Cole-Parmer (2808)
Product Type:
Availability:
In Stock (2883)
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 
Home > Cole-Parmer Catalog > search results for 'Palladium on carbon 10 50 wet with water for safety'

Showing 14925 results for 'Palladium on carbon 10 50 wet with water for safety'

Products
Technical Resources
 Product Detail
Sort
 Availability
 Price

Ruthenium on carbon, ca. 50% moisture 5% Ruthenium (25g)

  • Product Type: Chemical Reagents
  • Formula weight: 101.07
  • Empirical formula: Ru
EW-88208-18
Usually ships in 2 days.
$185.00 USD/EACH

Qty:

Palladium hydroxide on carbon powder 20% Pd, moisture ca 60% (10g)

  • Product Type: Chemical Reagents
  • CAS Number: 12135-22-7
  • Form: Powder
EW-88213-37
Usually ships in 2 days.
$260.00 USD/EACH

Qty:

Palladium on 4- to 8-mesh carbon 1% Pd (25g)

  • Product Type: Inorganic Chemical
  • Form: Solid
  • Formula weight: 106.42
EW-88207-65
Usually ships in 14 days.
$135.00 USD/EACH

Qty:

Cole-Parmer StableTemp Digital Utility Water Baths, 10 liters, 240V, 50 Hz

  • Product Type: Water Bath
  • Bath type: Utility Water Bath
  • Capacity: 10 Liters
EW-14576-10
In stock.
$563.00 USD/EACH

Qty:

Flowmeter, volumetric, for water, 0 to 10 mL/min

  • Product Type: Differential Pressure Flowmeter
  • Description: Differential Pressure Flowmeter
  • Max operating temperature: 122 º F
EW-32908-42
Usually ships in 13 days.
$1,601.00 USD/EACH

Qty:

Flowmeter, volumetric, for water, 0 to 10 L/min

  • Product Type: Differential Pressure Flowmeter
  • Description: Differential Pressure Flowmeter
  • Max operating temperature: 122 º F
EW-32908-49
Usually ships in 15 days.
$2,140.00 USD/EACH

Qty:

Safety Sign, Danger - Use Lockout Before Working On Equipment, 7" X 10", Plastic

  • Product Type: Signs
  • Material: Plastic
  • Model: MLKT021VP
EW-40710-91
Usually ships in 3 days.
$15.25 USD/EACH

Qty:

Reusable water filtration element with housing; 50 µm, 3/8" NPT(F)

  • Product Type: Water Filtration Elements
  • Housing dimensions: 7.375 in
  • Port size: 0.375 in NPT(F)
EW-29650-04
Usually ships in 7 days.
$152.00 USD/EACH

Qty:

RAE Systems gas detection tubes, carbon dioxide, 1 to 20%, 10/box

  • Product Type: Gas Detection Tubes
  • TLV: 5000 ppm
  • Gas: Carbon dioxide
EW-86260-57
In stock.
$35.00 USD/PACKAGE of 10

Qty:

polystat PPO Heated Bath, 5.3-liter, 100°C, 230V 50 Hz

  • Product Type: Heating Circulating Baths
  • Bath type: Open PPO Heated Water Bath
  • Capacity: 5.3 Liters
EW-12134-32
Usually ships in 10 days.
$1,296.00 USD/EACH

Qty:

Cole-Parmer Polystat Acrylic Heated Bath, 19-liter, 230V 50 Hz

  • Product Type: Heating Circulating Baths
  • Bath type: Open Acrylic Heated Water Bath
  • Capacity: 19 Liters
EW-12134-10
Usually ships in 10 days.
$1,602.00 USD/EACH

Qty:

Hach Nitrate Color Disc Test Kit, 10 ppm max, 50 tests/kit

  • Product Type: Water Quality Test Kit
  • Parameters: Nitrate
  • Description: Color wheel comparitive method
EW-05545-77
Usually ships in 14 days.
$106.00 USD/EACH
$100.70 USD/EACH

Qty:

polystat PPO Heated Bath, 21-liter, 100°C, 230V 50 Hz

  • Product Type: Heating Circulating Baths
  • Bath type: Open PPO Heated Water Bath
  • Capacity: 21 Liters
EW-12134-40
Usually ships in 10 days.
$1,569.00 USD/EACH

Qty:

Cole-Parmer Polystat Acrylic Heated Bath, 12.8-liter, 230V 50 Hz

  • Product Type: Heating Circulating Baths
  • Bath type: Open Acrylic Heated Water Bath
  • Capacity: 12.8 Liters
EW-12134-06
Usually ships in 10 days.
$1,580.00 USD/EACH

Qty:

Lab Companion Plant Growth Chamber with Humidity Control, 300 L, 230VAC, 50 Hz

  • Product Type: Plant Growth Chambers
  • Max temperature: 50 º C
  • Min temperature: Lights on: 10; lights off: 5 º C
EW-39353-15
Available for order
or call 1-888-358-4705 to order
$17,467.00 USD/EACH

Qty:

14925 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#