Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Search within results:
Go
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 
Home > Cole-Parmer Catalog > search results for 'Tool Kits'

Showing 20 results for 'Tool Kits'

Products
Technical Resources
 Product Detail
Sort
 Availability
 Price

Crescent 148 Piece Professional Tool Set

  • Product Type: Tool Sets
  • Qty/ea: 1
  • Manufacturer number: CTK148MP
EW-97105-64
In stock.
$191.00 USD/EACH

Qty:

Technician Starter Tool Kit; 37 Interchangeable Handles/Blades, Case

  • Product Type: Tool Sets
  • Dimensions: 13 1/2"W x 10 1/2"H x 2 3/8"D
  • Qty/ea: 1
EW-09900-72
In stock.
$517.00 USD/EACH

Qty:

Crescent 70 Piece Tool Set with Hard Case

  • Product Type: Tool Sets
  • Qty/ea: 1
  • Manufacturer number: CTK70MP
EW-97105-66
In stock.
$116.00 USD/EACH

Qty:

Dig Clinotronic W/Cal Kit Case 54-635-601

  • Product Type: Precison Measurement Tools
  • Qty/ea: 1
  • Brand: Fowler
EW-97604-15
In stock.
$1,778.00 USD/EACH

Qty:

Electronic caliper and data acquisition kit

  • Product Type: Measuring Kits/Sets
  • Range: 0 to 6 in/0 to 150 mm
  • Resolution: 0.0005 in/0.01 mm
EW-09923-38
In stock.
$306.00 USD/EACH

Qty:

EW-09254-20
In stock.
$43.00 USD/SET of 1

Qty:

Salisbury TK60 Hot Box 60-Piece Insulated Tool Kit

  • Product Type: Tool Sets
  • Qty/ea: 1
  • Manufacturer number: TK60
EW-91100-63
Usually ships in 4 days.
$3,429.00 USD/EACH

Qty:

Technician's Tool Kit, Polyethylene Case

  • Product Type: Tool Sets
  • Dimensions: 17 3/4"L x 12 5/8"H x 5 3/4"D
  • Qty/ea: 1
EW-09900-70
Available for order
or call 1-888-358-4705 to order
$1,086.00 USD/EACH

Qty:

Technician Tool Kit, Polyethylene Case with Wheels

  • Product Type: Tool Sets
  • Dimensions: 18"L x 15"H x 8"D
  • Qty/ea: 1
EW-09900-82
Available for order
or call 1-888-358-4705 to order
$1,241.00 USD/EACH

Qty:

Maintenance Tool Kit; 47 Piece Handle/Blades, Case, and Soldering Set

  • Product Type: Tool Sets
  • Dimensions: 17 3/4"L x 12 5/8"H x 4 3/4"D
  • Qty/ea: 1
EW-09900-74
Available for order
or call 1-888-358-4705 to order
$887.00 USD/EACH

Qty:

Gas Pressure Test Kit 0-5 PSI - 1.5% full-scale

  • Product Type: Pressure Test Kit
  • Wetted materials: Brass
  • Range: 0 to 5 psi
EW-97008-70
Usually ships in 5 days.
$45.00 USD/EACH
$40.50 USD/EACH

Qty:

Gass Pressure Test Kit 0-15 IN H2O - 1.5% full-scale

  • Product Type: Gas Test Gauges
  • Wetted materials: Brass
  • Range: 0 to 15 in. WC
EW-97008-71
Usually ships in 5 days.
$45.00 USD/EACH
$40.50 USD/EACH

Qty:

Borescope, 6" (152 mm)L, 0.039" (1.0 mm) dia

  • Product Type: Borescopes
  • Power: two C batteries (included)
  • Qty/ea: 1
EW-09744-64
Usually ships in 3 days.
$1,564.00 USD/EACH

Qty:

Miniature Laser Alignment Kits with Dot Image; Lens 41960-54, 50-125 ft

  • Product Type: Laser alignment tool
  • Range: 50 to 125 ft
  • Max output: <= 1 mW
EW-41935-22
Usually ships in 11 days.
$310.00 USD/EACH

Qty:

Borescope, 12" (305 mm) L, 0.039" (1.0 mm) dia

  • Product Type: Borescopes
  • Power: two C batteries (included)
  • Qty/ea: 1
EW-09744-66
Usually ships in 8 days.
$1,375.00 USD/EACH

Qty:

20 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#