log in or register
Price Range:
Availability:
Flask size (mL):
50 (1)
100 (1)
250 (1)
500 (1)
1000 (1)
2000 (1)
3000 (1)
5000 (1)
125 (1)
200 (1)
300 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
120 VAC models
120 VAC models
Hemispherical Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel supports (sold separately) are required to support mantle and allow attachment to ring stands. Supports have fabric strap basket to hold mantles;do not place mantle directly on ring stand.
Maximum temperature is 842°F (450°C). Mantles require a controlled electrical power source to prevent overheating. Order a temperature or a power controller below. Models for 120 VAC operation are CSA-certified.
 Product Detail
Sort
 Availability
 Price
Representative photo only Glas Col Flexible fabric heating mantle 1000 ml 380 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 1000-ml, 380 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 1000,Max temperature º F 842
EW-36225-40
In stock.
$219.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 100 ml 80 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 100-ml, 80 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 100,Max temperature º F 842
EW-36225-02
In stock.
$156.00 USD/EACH
Qty:
See all 120 VAC models
EW-36225-00 Glas-Col<small><sup>&reg;</sup></small> Flexible fabric heating mantle, 50-ml, 60 watts, 120 VAC
Glas-Col Flexible fabric heating mantle, 50-ml, 60 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 50,Max temperature º F 842
EW-36225-00
Usually ships in 9 days or sooner.
$141.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 250 ml 180 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 250-ml, 180 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 250,Max temperature º F 842
EW-36225-10
In stock.
$172.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 500 ml 270 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 500-ml, 270 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 500,Max temperature º F 842
EW-36225-30
In stock.
$192.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 5000 ml 600 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 5000-ml, 600 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 5000,Max temperature º F 842
EW-36225-70
Usually ships in 11 days.
$323.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 2000 ml 500 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 2000-ml, 500 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 2000,Max temperature º F 842
EW-36225-50
In stock.
$236.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 3000 ml 500 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 3000-ml, 500 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 3000,Max temperature º F 842
EW-36225-60
In stock.
$266.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 125 ml 80 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 125-ml, 80 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 125,Max temperature º F 842
EW-36225-04
Usually ships in 14 days.
$156.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 200 ml 100 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 200-ml, 100 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 200,Max temperature º F 842
EW-36225-06
Call 1-888-358-4705 for availability.
$160.00 USD/EACH
Qty:
See all 120 VAC models
Representative photo only Glas Col Flexible fabric heating mantle 300 ml 180 watts 120 VAC
Glas-Col Flexible fabric heating mantle, 300-ml, 180 watts, 120 VAC
Product Type Heating Mantle,Flask size mL 300,Max temperature º F 842
EW-36225-20
Call 1-888-358-4705 for availability.
$172.00 USD/EACH
Qty:
See all 120 VAC models
11 results (showing 1 - 11 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title