log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Metering Pumps > Solenoid/Electronic Diaphragm Pumps > Pulsafeeder Solenoid Metering Pumps > 4 to 20 mA Pace Pumps for High-Viscosity Applications
4 to 20 mA Pace Pumps for High-Viscosity Applications
4 to 20 mA Pace Pumps for High-Viscosity Applications
  • Reliable metering performance—guided dual check valves provide precise seating and excellent priming and suction lift
High-viscosity 4 to 20 mA pumps handle viscous fluids up to 20,000 cp. Pumps feature automatic control with 4 to 20 mA DC direct or external pacing, reliable timing circuits, stop function, and manually adjustable stroke rate and length. Operate pump in automatic or manual mode by selecting the proper setting on the "Auto-Off-Manual" switch. Circuit protection guards against voltage and current upsets. Install pumps indoors or outdoors.
 Product Detail
Sort
 Availability
 Price
EW-74147-10 High-Viscosity, 4 to 20 mA Pace Pump, 24 GPD, 115 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 24 GPD, 115 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 150 psi,Max flow rate LPH 3.76
EW-74147-10
Usually ships in 10 days.
$1,194.00 USD/EACH
Qty:
EW-74147-00 High-Viscosity, 4 to 20 mA Pace Pump, 12 GPD, 115 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 12 GPD, 115 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 150 psi,Max flow rate LPH 1.89
EW-74147-00
Usually ships in 14 days.
$939.00 USD/EACH
Qty:
EW-74147-20 High-Viscosity, 4 to 20 mA Pace Pump, 48 GPD, 115 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 48 GPD, 115 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 110 psi,Max flow rate LPH 7.57
EW-74147-20
Usually ships in 11 days.
$1,312.00 USD/EACH
Qty:
EW-74147-15 High-Viscosity, 4 to 20 mA Pace Pump, 24  GPD, 230 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 24 GPD, 230 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 150 psi,Max flow rate LPH 3.76
EW-74147-15
Usually ships in 14 days.
$1,105.00 USD/EACH
Qty:
EW-74147-30 High-Viscosity, 4 to 20 mA Pace Pump, 96 GPD, 115 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 96 GPD, 115 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 110 psi,Max flow rate LPH 15.14
EW-74147-30
Usually ships in 12 days.
$1,331.00 USD/EACH
Qty:
EW-74147-40 High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 115 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 115 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 80 psi,Max flow rate LPH 37.85
EW-74147-40
Usually ships in 10 days.
$1,550.00 USD/EACH
Qty:
EW-74147-05 High-Viscosity, 4 to 20 mA Pace Pump, 12 GPD, 230VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 12 GPD, 230VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 150 psi,Max flow rate LPH 1.89
EW-74147-05
Usually ships in 14 days.
$955.00 USD/EACH
Qty:
EW-74147-35 High-Viscosity, 4 to 20 mA Pace Pump, 96 GPD, 230 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 96 GPD, 230 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 110 psi,Max flow rate LPH 15.14
EW-74147-35
Usually ships in 14 days.
$1,351.00 USD/EACH
Qty:
EW-74147-25 High-Viscosity, 4 to 20 mA Pace Pump, 48 GPD, 230 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 48 GPD, 230 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 110 psi,Max flow rate LPH 7.57
EW-74147-25
Usually ships in 14 days.
$1,334.00 USD/EACH
Qty:
EW-74147-45 High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 230 VAC, 50/60 Hz.  Representative Photo Only
High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 230 VAC, 50/60 Hz
Product Type Diaphragm Metering Pumps,Max pressure 80 psi,Max flow rate LPH 37.85
EW-74147-45
Usually ships in 13 days.
$1,571.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Pumps > Metering Pumps > Solenoid/Electronic Diaphragm Pumps > Pulsafeeder Solenoid Metering Pumps > 4 to 20 mA Pace Pumps for High-Viscosity Applications
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title