Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

AIM600 Programmable Digestion Systems

AIM600 Programmable Digestion Systems

  • Increase productivity with programmable digestions and large vial capacity
  • Perform digestions of aqueous samples for metals analysis by Atomic Absorption (AA) or Inductively Coupled Plasma (ICP)
  • Conduct Kjeldahl digestion of food products or plant tissue for nitrogen & phosphorous analysis
  • Free up valuable time by digesting large numbers of samples—up to 50 vials—simultaneously
Eliminate the need for repetitive small batch digestions with these compact block digestion systems. These systems can handle sample batches of 28 or 50 at a time. Run your digest overnight and have a full batch of digested samples ready for analysis at the start of the next day. The temperature of each digestion position is tightly regulated and controlled, ensuring that all the samples in each batch are digested equally. Store up to nine different programs, each with up to forty steps, for automating the most complex digestion procedures, allowing reproducibility from batch to batch, operator to operator. Controlled heating rates, as low as 1°C per minute, protect against the bumping and splattering of reactive samples.

A built in safety cut-out feature within the controller is triggered automatically if the programmed temperature is exceeded by 30°C or if the maximum set temperature is exceeded. The controller is separate from the digestion block, protecting the electronics and the operator from any digest acid fumes. Apart from the power and reagents, these block digestion systems have no ongoing
consumable costs and do not use any limited life components.
 Product Detail
Sort
 Availability
 Price
Representative photo only AIM600 Programmable Block Digestion System 28 Place With 250 mL Tubes

AIM600 Programmable Block Digestion System, 28 Place With 250 mL Tubes

Product Type Programmable Block Digestion System,Max operating temperature º F 842,Max operating temperature º C 450
EW-78871-51
Call 1-888-358-4705 for availability.
$8,360.00 USD/EACH

Qty:

Representative photo only Tubes Volumetric 250 mL 28 pk
EW-78870-80
Usually ships in 2 days.
$811.50 USD/PACKAGE

Qty:

Representative photo only Tubes Volumetric 75 mL 50 pk
EW-78870-72
Usually ships in 13 days.
$1,026.00 USD/PACKAGE

Qty:

Representative photo only AIM600 Programmable Block Digestion System 50 Place With 75 mL Tubes

AIM600 Programmable Block Digestion System, 50 Place With 75 mL Tubes

Product Type Programmable Block Digestion System,Max operating temperature º F 842,Max operating temperature º C 450
EW-78871-54
Usually ships in 10 days.
$8,360.00 USD/EACH

Qty:

Representative photo only Tubes Straight 250 mL 28 pk
EW-78870-78
Usually ships in 2 days.
$794.00 USD/PACKAGE

Qty:

Representative photo only Tubes Straight 75 mL 50 pk
EW-78870-70
Usually ships in 2 days.
$819.00 USD/PACKAGE

Qty:

Representative photo only Tubes Volumetric 100 mL 50 pk
EW-78870-76
Usually ships in 2 days.
$1,036.00 USD/PACKAGE

Qty:

Representative photo only AIM600 Programmable Block Digestion System 50 Place With 100 mL Tubes

AIM600 Programmable Block Digestion System, 50 Place With 100 mL Tubes

Product Type Programmable Block Digestion System,Max operating temperature º F 842,Max operating temperature º C 450
EW-78871-56
Usually ships in 7 days.
$8,360.00 USD/EACH

Qty:

8 results (showing 1 - 8 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title