log in or register
Product Type:
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Air Compressors > Accessories for Compressors - Air Equipment > ARO-Flo Series Filters, Regulators, and Lubricators
ARO-Flo Series Filters, Regulators, and Lubricators
ARO-Flo Series Filters, Regulators, and Lubricators
FILTERS
ARO-Flo compressed air filters are designed to remove airborne solid and liquid contaminants. Filters can be ordered with different elements, including coalescing models which are capable of removing oil aerosols and particles down to 0.3 micron.
REGULATORS
Air line regulators provide controlled, consistent air pressure as required for specific pneumatic equipment connected to the air system. All ARO-Flo regulators are offered with a standard adjustment range of 0 – 140 psig (0 – 9.6 barg).
LUBRICATORS
ARO-Flo mist-type lubricators help ensure that pneumatic devices receive the required lubrication to maintain peak performance, reduce wear, and prolong service life. Precise oil feed adjustment sets the proper oil drip rate.
NOTE: Combinations are available online at the www.coleparmer.com/AODD
 Product Detail
Sort
 Availability
 Price
Representative photo only Mini Coalescing Air Filter 0 3 micron coalescing filter Max 49 SCFM Max Pressure 150 PSIG 1 4 NPT F connection
Mini Coalescing Air Filter, 0.3 micron coalescing filter, Max 49 SCFM, Max Pressure 150 PSIG, 1/4" NPT(F) connection
Product Type Compressor Filters/Regulators,Max pressure psi 150,Brand ARO-Flo
EW-75300-77
In stock.
$53.50 USD/EACH
Qty:
Representative photo only HD Air Filter 5 micron filter Max 288 SCFM Max Pressure 250 PSIG 3 4 NPT F connection
HD Air Filter, 5 micron filter, Max 288 SCFM, Max Pressure 250 PSIG, 3/4" NPT(F) connection
Product Type Compressor Filters/Regulators,Max pressure psi 250,Brand ARO-Flo
EW-75300-84
Usually ships in 13 days.
$156.00 USD/EACH
Qty:
Representative photo only Mid Size Air Filter 5 micron filter Max 197 SCFM Max Pressure 150 PSIG 1 2 NPT F connection
Mid Size Air Filter, 5 micron filter, Max 197 SCFM, Max Pressure 150 PSIG, 1/2" NPT(F) connection
Product Type Compressor Filters/Regulators,Max pressure psi 150,Brand ARO-Flo
EW-75300-83
In stock.
$73.50 USD/EACH
Qty:
Representative photo only Combo Filter and Regulator and Lubricator Max 61 SCFM Max Pressure of 150 PSIG Temperature Range of 23 125 F 5 Micron Filter 1 4 NPT F connection with a Polycarbonate Bowl and manual valve
EW-75300-71
Usually ships in 13 days.
$203.00 USD/EACH
Qty:
Representative photo only Compac Coalescing Air Filter 0 3 micron coalescing filter Max 73 SCFM Max Pressure 150 PSIG 1 4 NPT F connection
Compac Coalescing Air Filter, 0.3 micron coalescing filter, Max 73 SCFM, Max Pressure 150 PSIG, 1/4" NPT(F) connection
Product Type Compressor Filters/Regulators,Max pressure psi 150,Brand ARO-Flo
EW-75300-79
Usually ships in 13 days.
$74.00 USD/EACH
Qty:
EW-75300-46 Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4" NPT(F) connection
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4" NPT(F) connection
Product Type Compressor Regulators,Max pressure psi 0 to 140,Brand ARO-Flo
EW-75300-46
Usually ships in 13 days.
$26.00 USD/EACH
Qty:
EW-75300-67 Compact Lubricator with bowl guard, Max 105 SCFM, Max Pressure of 150 PSI, 3/8" NPT(F) connection
Compact Lubricator with bowl guard, Max 105 SCFM, Max Pressure of 150 PSI, 3/8" NPT(F) connection
Product Type Compressor Lubricators,Max pressure psi 150,Brand ARO-Flo
EW-75300-67
Usually ships in 13 days.
$69.00 USD/EACH
Qty:
Representative photo only Mini Air Regulator Max 45 SCFM Pressure range of 0 140 PSIG 1 8 NPT F connection
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8" NPT(F) connection
Product Type Compressor Regulators,Max pressure psi 0 to 140,Brand ARO-Flo
EW-75300-45
Usually ships in 13 days.
$26.00 USD/EACH
Qty:
Representative photo only Mid Size Lubricator with bowl guard Max 156 SCFM Max Pressure of 150 PSI 1 2 NPT F connection
Mid Size Lubricator with bowl guard, Max 156 SCFM, Max Pressure of 150 PSI, 1/2" NPT(F) connection
Product Type Compressor Lubricators,Max pressure psi 150,Brand ARO-Flo
EW-75300-69
Usually ships in 13 days.
$81.00 USD/EACH
Qty:
Representative photo only Mid Size Air Regulator Max 215 SCFM Pressure range of 0 140 PSIG 1 2 NPT F connection
Mid Size Air Regulator, Max 215 SCFM, Pressure range of 0 - 140 PSIG, 1/2" NPT(F) connection
Product Type Compressor Regulators,Max pressure psi 0 to 140,Brand ARO-Flo
EW-75300-49
Usually ships in 13 days.
$57.00 USD/EACH
Qty:
Representative photo only Combo Filter Regulator and Lubricator Max 96 SCFM Max Pressure of 150 PSIG Temperature Range of 23 125 F 5 Micron Filter 3 8 NPT F connection with a Polycarbonate Bowl and manual valve
EW-75300-73
Usually ships in 13 days.
$203.00 USD/EACH
Qty:
Representative photo only Mini Air Regulator Max 47 SCFM 5 Micron Filter Pressure range of 0 140 PSIG 1 4 NPT F connection
Mini Air Regulator, Max 47 SCFM, 5 Micron Filter, Pressure range of 0 - 140 PSIG, 1/4" NPT(F) connection
Product Type Compressor Filters/Regulators,Max pressure psi 0 to 140,Brand ARO-Flo
EW-75300-57
Usually ships in 13 days.
$50.00 USD/EACH
Qty:
Representative photo only Mid Size Coalescing Air Filter 0 3 micron coalescing filter Max 156 SCFM Max Pressure 150 PSIG 3 8 NPT F connection
EW-75300-82
Usually ships in 13 days.
$110.00 USD/EACH
Qty:
Representative photo only Mini Lubricator with bowl guard Max 32 SCFM Max Pressure of 150 PSI 1 8 NPT F connection
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI, 1/8" NPT(F) connection
Product Type Compressor Lubricators,Max pressure psi 150,Brand ARO-Flo
EW-75300-66
Usually ships in 13 days.
$47.50 USD/EACH
Qty:
Representative photo only Combo Filter Regulator and Lubricator Max 44 SCFM Max Pressure of 150 PSIG Temperature Range of 23 125 F 5 Micron Filter 1 4 NPT F connection with a Polycarbonate Bowl and manual valve
EW-75300-70
Usually ships in 13 days.
$112.00 USD/EACH
Qty:
26 results (showing 1 - 15 )
Per Page 
Home  >  Pumps > Air Compressors > Accessories for Compressors - Air Equipment > ARO-Flo Series Filters, Regulators, and Lubricators
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title