log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Metal Detectors > Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems > Accessories for Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems
Accessories for Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems
Choose from our comprehensive line of Accessories for Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems and other Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific AuditCheck Performance Verification System
Thermo Scientific AuditCheck Performance Verification System
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-28
Call 1-888-358-4705 for availability.
$750.00 USD/EACH
Qty:
Representative photo only Thermo Scientific High Polish Finish for Head and Rejector only for APEX 500 Rx
Thermo Scientific High Polish Finish for Head and Rejector only for APEX 500 Rx
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-29
Call 1-888-358-4705 for availability.
$1,500.00 USD/EACH
Qty:
Representative photo only Thermo Scientific IQ OQ PQ Validation Documentation
Thermo Scientific IQ/OQ/PQ Validation Documentation
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-33
Call 1-888-358-4705 for availability.
$4,000.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Rejector with 2 inch 50 8 mm OD for hose attachement for reject product flow replace bin
EW-54502-32
Call 1-888-358-4705 for availability.
$650.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Rejector with 2 inch 50 8 mm OD for hose attachement for good product flow
Thermo Scientific Rejector with 2 inch (50.8 mm) OD for hose attachement for good product flow
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-31
Call 1-888-358-4705 for availability.
$650.00 USD/EACH
Qty:
Representative photo only Thermo Scientific High Polish Finish for complete system APEX 500 Rx
Thermo Scientific High Polish Finish for complete system APEX 500 Rx
Product Type Accessory,CE Compliance Yes,Brand Thermo Scientific
EW-54502-30
Call 1-888-358-4705 for availability.
$2,500.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
Home  >  Safety Products > Metal Detectors > Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems > Accessories for Thermo Scientific APEX 500 Rx Pharmaceutical Metal Detection Systems
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title