United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Brand:
Cole-Parmer
(8)
Product Type:
Chemical Reagents
(65)
Chemical Dyes
(1)
Price Range:
$0 to $100
(29)
$100 to $299
(32)
$300 to $499
(2)
[+]
Show more
(3)
$500 to $799
(3)
$1,100 to $1,999
(1)
$2,000 and Over
(1)
[-]
Show less
Availability:
In Stock
(56)
Form:
Liquid
(64)
Quantity (mL):
4000
(19)
1000
(15)
500
(7)
[+]
Show more
(10)
2500
(7)
20000
(4)
7500
(4)
200000
(2)
100
(1)
3785
(1)
10
(1)
50
(1)
950
(1)
25000
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Chemicals, Reagents, & Lubricants
>
Chemicals
>
Chemicals A
>
Aceto to Acetyle
> Acetone
Acetone
Choose from our comprehensive line of Acetone and other Aceto to Acetyle products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Acetone (Electronic) (4 liter)
Product Type Chemical Reagents,Purity grade Electronic,Form Liquid
EW-88000-66
In stock.
$220.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone Acs 4L
Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-08
Usually ships in 5 days.
$81.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (Histological) (4 liter) poly
Product Type Chemical Reagents,Purity grade Histological,Form Liquid
EW-88000-78
In stock.
$130.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone, reagent ACS 99.5% (4l)
Product Type Chemical Reagents,Form Liquid,Hazard symbol Highly_Flammable, Irritant
EW-88360-49
In stock.
$175.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (For High-resolution Chromatography Suitable for Organic Residue Analysis/GC Resolv*) (4 liter)
Product Type Chemical Dyes,Purity grade GC Resolv
,Form Liquid
EW-88000-50
In stock.
$260.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (HPLC) (4 liter)
Product Type Chemical Reagents,Purity grade HPLC,Form Liquid
EW-88000-60
In stock.
$193.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone Hplc 4L
Product Type Chemical Reagents,Purity grade HPLC,Form Liquid
EW-89000-11
Usually ships in 4 days.
$130.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (HPLC) (1 liter)
Product Type Chemical Reagents,Purity grade HPLC,Form Liquid
EW-88000-56
In stock.
$94.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone CP 4L
Product Type Chemical Reagents,Purity grade CP,Form Liquid
EW-89000-13
Usually ships in 4 days.
$100.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone 99+% (2.5l)
Product Type Chemical Reagents,Form Liquid,Hazard symbol Highly_Flammable, Irritant
EW-88190-15
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone Certified Acs 20l
Product Type Chemical Reagents,Form Liquid,Quantity mL 20000
EW-88050-37
In stock.
$443.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone Acs 1L
Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-07
Usually ships in 5 days.
$39.50 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (Certified ACS) (4 liter)
Product Type Chemical Reagents,Purity grade Certified ACS,Form Liquid
EW-88000-72
In stock.
$190.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone (Histological) (1 gal.)
Product Type Chemical Reagents,Purity grade Histological,Form Liquid
EW-88000-82
In stock.
$110.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
Acetone ACS 500mL
Product Type Chemical Reagents,Purity grade ACS,Form Liquid
EW-89000-06
Usually ships in 6 days.
$14.50 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Acetone
66
results (showing
1
-
15
)
1
2
3
4
5
Next >
>>
Per Page
5
10
15
25
50
Home
>
Chemicals, Reagents, & Lubricants
>
Chemicals
>
Chemicals A
>
Aceto to Acetyle
> Acetone
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title