Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Acridine Orange Hemizinc Salt

Acridine Orange Hemizinc Salt

Acridine Orange Hemizinc Salt

  • Solubility - To pass test
  • Loss on Drying at 105°C - ≤5%
  • E1%1cm - ≥1100L g-1cm-1
  • Lambda Max. (λ) - 490nm ±2nm
 Product Detail
Sort
 Availability
 Price
Representative photo only Acridine Orange Hemizinc Salt 10g

Acridine Orange Hemizinc Salt (10g)

Product Type Chemical Reagents,Hazard symbol Harmful
EW-88056-57
In stock.
$83.00 USD/EACH

Qty:

1 results (showing 1 - 1 )
Per Page 
Home  >  Acridine Orange Hemizinc Salt

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title