log in or register
Price Range:
Readability (g):
0.01 (5)
0.001 (4)
Capacity (g):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Balances & Scales > Toploading Balances > Adam Equipment Toploading Balances > Adam Equipment PGW Precision Toploading Balances
Adam Equipment PGW Precision Toploading Balances
Adam Equipment PGW Precision Toploading Balances
  • Improved response times and enhanced stability and features
  • GLP-compliant printouts via RS-232 connection
  • Digital filters compensate for vibration, drafts or temperature changes
  • Parts counting, check weighing, percent weighing, animal weighing
  • Below-balance weighing hook
  • Date and time stored in memory
  • Oversized backlit LCD
These balances feature multiple weighing modes including parts counting, check weighing, animal weighing, net/total formulation, liquid density, solid density, and percentage weighing. An oversized backlit LCD with split segments and 1” high digits offers easy visibility. The capacity tracker graphically displays percentages and notifies you of any approaching overloads. All models feature an RS-232 port for configuration to a printer—providing GLP-compliant reports after each calibration.
A second output port may be linked to other devices such as USB, Ethernet, or a remote display whenever additional communication outputs are desired. The balances feature 17 weighing units including one custom unit and seven weighing modes. Toploading balances offer choice of internal or external calibration; deactivate internal calibration at any time to perform external calibrations.
 Product Detail
Sort
 Availability
 Price
EW-11701-41 Representative Photo Only.  Adam PGW Precision Toploading Balance, 450 g x 0.001 g - 115 VAC
Adam PGW Precision Toploading Balance, 450 g x 0.001 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 450
EW-11701-41
Usually ships in 14 days.
$1,075.00 USD/EACH
Qty:
EW-11701-42 Representative Photo Only.  Adam PGW Precision Toploading Balance, 750 g x 0.001 g  - 115 VAC
Adam PGW Precision Toploading Balance, 750 g x 0.001 g- 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 750
EW-11701-42
Usually ships in 14 days.
$1,475.00 USD/EACH
Qty:
EW-11701-39 Representative Photo Only.  Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 150
EW-11701-39
Usually ships in 14 days.
$875.00 USD/EACH
Qty:
EW-11701-43 Representative Photo Only.  Adam PGW Precision Toploading Balance, 1500 g x 0.01 g - 115 VAC
Adam PGW Precision Toploading Balance, 1500 g x 0.01 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 1500
EW-11701-43
Usually ships in 14 days.
$875.00 USD/EACH
Qty:
EW-11701-45 Representative Photo Only.  Adam PGW Precision Toploading Balance, 3500 g x 0.01 g - 115 VAC
Adam PGW Precision Toploading Balance, 3500 g x 0.01 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 3500
EW-11701-45
Usually ships in 14 days.
$1,075.00 USD/EACH
Qty:
EW-11701-44 Representative Photo Only.  Adam PGW Precision Toploading Balance, 2500 g x 0.01 g - 115 VAC
Adam PGW Precision Toploading Balance, 2500 g x 0.01 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 2500
EW-11701-44
Usually ships in 14 days.
$975.00 USD/EACH
Qty:
EW-11701-56 Representative Photo Only.  Adam PGW Precision Toploading Balance, 4500 g x 0.01 g - Internal Calibration - 115 VAC
Adam PGW Precision Toploading Balance, 4500 g x 0.01 g - Internal Calibration - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 4500
EW-11701-56
Usually ships in 14 days.
$1,425.00 USD/EACH
Qty:
EW-11701-40 Representative Photo Only.  Adam PGW Precision Toploading Balance, 250 g x 0.001 g - 115 VAC
Adam PGW Precision Toploading Balance, 250 g x 0.001 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 250
EW-11701-40
Usually ships in 14 days.
$975.00 USD/EACH
Qty:
EW-11701-46 Representative Photo Only.  Adam PGW Precision Toploading Balance, 4500 g x 0.01 g - 115 VAC
Adam PGW Precision Toploading Balance, 4500 g x 0.01 g - 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 4500
EW-11701-46
Usually ships in 14 days.
$1,175.00 USD/EACH
Qty:
9 results (showing 1 - 9 )
Per Page 
Home  >  Balances & Scales > Toploading Balances > Adam Equipment Toploading Balances > Adam Equipment PGW Precision Toploading Balances
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title