Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Brand:
Availability:
Quantity (g):
500 (4)
100 (1)
2000 (1)
Form:
Powder (3)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Agar, Granular

Agar, Granular
Choose from our comprehensive line of Agar and other Ae to Allylh products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Agar 500g

Agar 500g

Product Type Agar,Purity grade Purified,Form Powder
EW-89000-20
Usually ships in 7 days.
$95.00 USD/EACH

Qty:
See all Agar, Granular

Representative photo only Agar Granulated Molecular Genetics 500g

Agar (Granulated/Molecular Genetics) (500g)

Product Type Agar,Purity grade Molecular Genetics,Form Granular
EW-88057-36
In stock.
$165.00 USD/EACH

Qty:
See all Agar, Granular

Representative photo only Agar USP NF 80 100 Mesh 100g

Agar (USP/NF, 80-100 Mesh) (100g)

Product Type Agar,Purity grade USP/NF,Form Powder
EW-88061-31
In stock.
$66.00 USD/EACH

Qty:
See all Agar, Granular

Representative photo only Agar USP NF 80 100 Mesh 500g

Agar (USP/NF, 80-100 Mesh) (500g)

Product Type Agar,Purity grade USP/NF,Form Powder
EW-88061-32
Usually ships in 7 days.
$239.00 USD/EACH

Qty:
See all Agar, Granular

Representative photo only Agar Granulated Molecular Genetics 2kg

Agar (Granulated/Molecular Genetics) (2kg)

Product Type Agar,Purity grade Molecular Genetics,Form Granular
EW-88057-35
In stock.
$599.00 USD/EACH

Qty:
See all Agar, Granular

Representative photo only Agar Laboratory 500g

Agar (Laboratory) (500g)

Product Type Agar,Purity grade Laboratory,Form Granular / Powder
EW-88001-13
In stock.
$321.00 USD/EACH

Qty:
See all Agar, Granular

6 results (showing 1 - 6 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title