Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Chemicals, Reagents, & Lubricants > Agar and Agarose

Agar and Agarose

Agar and Agarose

Agar is a versatile substance that is found in everything from Japanese desserts to microbiology cultures. In the lab agar is purified and heated to create aragose which is used in electrophoresis. Agar culture media is commonly used for isolation, differentiation, identification, susceptibility, and quality control. There are a variety of media formulations and packaging options: including dehydrated culture media, prepared media bottles and bags, and finally electrophoresis gels. Choose from pre-poured plates to save you time, or dehydrated agar powder and media for longer storage times and to save on costs. Having trouble choosing the right type of culture media based on your application? Cole-Parmer can offer assistance with your agar selection or with any custom ordering needs.
 Product Detail
Sort
 Availability
 Price
EW-14207-70 Representative image only.  Check individual details.

Thermo Scientific Contact plate sterile tryptic soy agar with lecithin, polysorbate 80, 10/PK

Product Type Prepared Media Plates,Description Irradiated. For cultivation of bacteria, yeast, and fungi.,Media Contact plate sterile tryptic soy agar with lecithin, polysorbate 80,Qty/pk 10
EW-14207-70
Usually ships in 4 days.
$42.75 USD/PACKAGE

Qty:

EW-14207-00 Representative image only.  Check individual details.

Thermo Scientific Tryptic soy agar (TSA), soybean casein digest, 10/pk

Product Type Prepared Media Plates,Description General-purpose medium for cultivation and isolation of microorganisms. Industrial reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW,Media Tryptic soy agar (TSA), soybean casein digest,Qty/pk 10
EW-14207-00
Usually ships in 8 days.
$8.05 USD/PACKAGE

Qty:

EW-14200-28 Representative Image. Check individual details.

Cole-Parmer Potato dextrose agar, 500 g jar

Product Type Agar,Capacity g 500,Qty/pk 1
EW-14200-28
Usually ships in 4 days.
$69.00 USD/EACH

Qty:

EW-99532-44 BioPaddles<small><sup>&#153;</sup></small>

LaMotte BioPaddles Sabouraud Dextrose Agar Microbiological Test Kit

Product Type Microbiological,Description Test kit,Brand LaMotte
EW-99532-44
Call 1-888-358-4705 for availability.
$39.95 USD/EACH

Qty:
See all

EW-99532-20 BioPaddles<small><sup>&#153;</sup></small>

LaMotte BioPaddles R2A/MacConkey Agar Microbiological Test Kit

Product Type Microbiological,Description Test kit,Brand LaMotte
EW-99532-20
In stock.
$40.00 USD/EACH

Qty:
See all

Representative photo only Agar 500g

Agar 500g

Product Type Agar,Purity grade Purified,Form Powder
EW-89000-20
Usually ships in 7 days.
$95.00 USD/EACH

Qty:
See all Agar, Granular

EW-14200-12 Representative Image. Check individual details.

Cole-Parmer Brain heart infusion agar, 500 g jar

Product Type Agar,Capacity g 500,Qty/pk 1
EW-14200-12
Usually ships in 4 days.
$114.00 USD/EACH

Qty:

EW-14200-34 Cole-Parmer Tryptic Soy Agar, 500 g jar

Cole-Parmer Tryptic Soy Agar, 500 g jar

Product Type Agar,Capacity g 500,Qty/pk 1
EW-14200-34
Usually ships in 7 days.
$68.00 USD/EACH

Qty:

EW-99532-22 BioPaddles<small><sup>&#153;</sup></small>

LaMotte BioPaddles Nut-TTC/EMB Agar Microbiological Test Kit

Product Type Microbiological,Description Test kit,Brand LaMotte
EW-99532-22
In stock.
$40.00 USD/EACH

Qty:
See all

EW-14202-62 Representative Image. Check individual details.

Cole-Parmer Sabouraud Dextrose Agar, 500 g

Product Type Agar,Capacity g 500,Qty/pk 1
EW-14202-62
Usually ships in 6 days.
$56.00 USD/EACH

Qty:

EW-14206-72 Representative image only.  Check individual details.

Thermo Scientific Blood agar (TSA with 5% sheep blood), 10/pk

Product Type Prepared Media Plates,Description General-purpose medium for cultivation and isolation of microorganisms. Industrial reference: BAM,Media Blood agar (TSA with 5% sheep blood),Qty/pk 10
EW-14206-72
Usually ships in 6 days.
$7.50 USD/PACKAGE

Qty:

EW-14207-92 Representative image only.  Check individual details.

Thermo Scientific Potato dextrose agar, pH 5.6, 10/pk

Product Type Prepared Media Plates,Description For cultivation, identification, and enumeration of yeast and fungi. Industrial reference: BAM, COMPF, USP,Media Potato dextrose agar, pH 5.6,Qty/pk 10
EW-14207-92
Usually ships in 11 days.
$10.55 USD/PACKAGE

Qty:

EW-99532-42 BioPaddles<small><sup>&#153;</sup></small>

LaMotte BioPaddles Nutrient Agar Microbiological Test Kit

Product Type Microbiological,Description Test kit,Brand LaMotte
EW-99532-42
Usually ships in 8 days.
$39.95 USD/EACH

Qty:
See all

Representative photo only Agar Granulated Molecular Genetics 500g

Agar (Granulated/Molecular Genetics) (500g)

Product Type Agar,Purity grade Molecular Genetics,Form Granular
EW-88057-36
In stock.
$165.00 USD/EACH

Qty:
See all Agar, Granular

EW-14200-22 Representative Image. Check individual details.

Cole-Parmer MacConkey agar, 500 g jar

Product Type Agar,Capacity g 500,Qty/pk 1
EW-14200-22
Usually ships in 7 days.
$75.00 USD/EACH

Qty:
See all MacConkey Agar

75 results (showing 1 - 15 )
Per Page 
Home  >  Chemicals, Reagents, & Lubricants > Agar and Agarose

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title