United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$1,100 to $1,999
(1)
$2,000 and Over
(1)
Max flow rate (LPM):
68.1
(1)
98.4
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Positive Displacement Pumps
>
Air-Driven Pumps
> Air-Operated Powder Pumps
Air-Operated Powder Pumps
Transfer fine-grained (100 µm or finer), low bulk density dry powders in a dust-free environment
Aluminum or 316 SS housings
Patented non-lubricated, non-stalling air valve technology
Compressed air induction system fluidizes all four check valves
Air-Driven Pumps and other Pumps - Positive Displacement Pumps products are now available to order from the Cole-Parmer Catalog. Click any Air-Driven Pumps category to view all models and products.
Eliminate the need for unsafe and labor intensive augers and conveyors for moving bulk powders. These powder pumps are heavy duty and a more cost-effective method to move bulk solids effectively through your process. Pump particles that are 150 mesh (106 micron) or smaller. Pumps handle bulk densities up to 50 lbs/ft
3
. Pumps can be located up to a maximum of 15 feet above the powder source.
Note:
Pumps will convey at different distances depending upon the micron size and bulk density of the powder. Contact our Application Specialists for more information about conveying distances for specific materials.
What's included:
conduit box for hard-wiring.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Air-operated powder pump, 18 GPM, 316 SS, Santoprene, EPDM
Product Type Powder Pumps,Max flow rate LPM 98.4,Duty cycle Continuous
EW-74026-10
Usually ships in 7 days.
$3,159.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Air-Operated Powder Pumps
Air-operated powder pump, 18 GPM, Aluminum, Santoprene, EPDM
Product Type Powder Pumps,Max flow rate LPM 68.1,Duty cycle Continuous
EW-74026-00
Usually ships in 10 days.
$1,690.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Air-Operated Powder Pumps
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Positive Displacement Pumps
>
Air-Driven Pumps
> Air-Operated Powder Pumps
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title