Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Ammonium Hydroxide

Ammonium Hydroxide
Choose from our comprehensive line of Ammonium Hydroxide and other Ammonium products.
 Product Detail
Sort
 Availability
 Price

Cole-Parmer Ammonium Hydroxide, ACS Grade, 2.5 L

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 1336-21-6
EW-88401-67
Available for order
or call 1-888-358-4705 to order
$69.00 USD/EACH

Qty:

Ammonium Hydroxide, 50% (v/v) Aqueous Solution (1 + 1) (1 gal)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86984-82
Available for order
or call 1-888-358-4705 to order
$126.00 USD/EACH

Qty:

Cole-Parmer Ammonium Hydroxide, ACS Grade, 1 L

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 1336-21-6
EW-88401-66
Available for order
or call 1-888-358-4705 to order
$38.50 USD/EACH

Qty:

Ammonium Hydroxide (26°Bé/NF/FCC) (2.5 liter)

  • Product Type: Inorganic Chemical
  • Purity grade: NF or FCC
  • CAS Number: Includes 1336216 and 7732185
EW-88001-84
Usually ships in 4 days.
$203.00 USD/EACH

Qty:

Ammonium Hydroxide, 5% (v/v) Aqueous Solution (1 + 19) (20 L)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86992-94
Usually ships in 14 days.
$143.00 USD/EACH

Qty:

Ammonium Hydroxide, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86969-70
Available for order
or call 1-888-358-4705 to order
$39.25 USD/EACH

Qty:

Ammonium Hydroxide, 20% (v/v) Aqueous Solution (1 + 4) (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86969-69
Available for order
or call 1-888-358-4705 to order
$37.00 USD/EACH

Qty:

Cole-Parmer Ammonium Hydroxide, ACS Grade, 500 mL

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 1336-21-6
EW-88401-68
Available for order
or call 1-888-358-4705 to order
$38.25 USD/EACH

Qty:

Ammonium Hydroxide 25% 2.5lt

  • Product Type: Chemical Reagents
  • Form: Liquid
  • Quantity: 2500 mL
EW-88252-97
Usually ships in 14 days.
$115.00 USD/EACH

Qty:

Ammonium Hydroxide, 10% (v/v) Aqueous Solution (1 + 9) (1 gal)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86984-79
Usually ships in 7 days.
$67.25 USD/EACH

Qty:

Ammonium Hydroxide (Certified ACS Plus) (500mL)

  • Product Type: Chemical Reagent
  • Purity grade: Certified ACS Plus
  • CAS Number: 1336-21-6
EW-88001-78
Usually ships in 2 days.
$109.00 USD/EACH

Qty:

Ammonium Hydroxide, 5.00 Normal (1 gal) - Liquid

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 5.00
EW-86984-84
Available for order
or call 1-888-358-4705 to order
$155.00 USD/EACH

Qty:

Ammonium Hydroxide, 16.7% (v/v) Aqueous Solution (1 + 5), Electronic Grade (32 oz)

  • Product Type: Inorganic Chemical
  • Purity grade: Electronic grade
  • CAS Number: Includes 1336-21-6 and 7732-18-5
EW-86977-94
Available for order
or call 1-888-358-4705 to order
$52.75 USD/EACH

Qty:

Ammonium Hydroxide, 2.00 Normal (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 2.00
EW-86969-72
Usually ships in 9 days.
$33.50 USD/EACH

Qty:

Ammonium Hydroxide, 5.00 Normal (32 oz) - Liquid

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 5.00
EW-86978-01
Available for order
or call 1-888-358-4705 to order
$64.00 USD/EACH

Qty:

43 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#