United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Capacity (cu ft):
2
(1)
4
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Heating Equipment
>
Sterilizers
>
Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
> Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
Gently sterilize delicate items that heat and moisture can damage
Sterilize any medical instrument, component, or material
Sterilizer requires less than 17.5 grams of Ethylene Oxide per cycle
Sterilize and aerate in the same cabinet
These gas sterilizer systems use 100% Ethylene Oxide (EtO) to safely sterilize delicate items that heat and moisture can damage. EtO sterilizers operate at room temperature and atmospheric pressure—safe for electronics, fiber optics, cameras, and plastic or rubber items such as tubing. Sterilizers feature a combination of ventilation and purge systems ensuring that EtO exposure levels are below OSHA prescribed limits. The advanced purge system allows for items to be aerated in the sterilization cabinet for two hours at the end of a cycle and will continue indefinitely until items are removed.
Sterilizers use unique plastic liner bags and individual gas ampoules. Two sizes of bag/ampoule combinations are available to match the size of the load—never use more gas than you need to. Since sterilizer systems use only small amounts of EtO gas, they meet all EPA requirements without the need for expensive abatement equipment. A dedicated exhaust line to the outside for the outlet hose is all that is required. Order outlet hose separately under “Accessories” at right. Sterilizer systems include everything necessary for the sterilization process, including biological and chemical indicators for verifying the sterility and EtO exposure of items. Sterilizers are available in two sizes—choose the
long model for handling larger items.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Standard sterilizer system, 2.0 cu ft 115 VAC, 60 Hz
Product Type Sterilizer,Capacity cu ft 2,Chamber size " W 13
EW-10778-00
Usually ships in 4 days.
$5,350.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
Long Sterilizer System, 4.0 cu ft 115 VAC, 60HZ
Product Type Sterilizer,Capacity cu ft 4,Chamber size " W 18
EW-10778-20
Usually ships in 5 days.
$7,750.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Heating Equipment
>
Sterilizers
>
Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
> Anprolene Ethylene Oxide (EtO) Gas Sterilizer Systems
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title