United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Ansell Vinyl Coat Apron
(5)
Cryo-Apron Aprons
(3)
Aluminized Zetex Apron
(3)
Ansell Endurosaf Aprons
(3)
Ansell Neoprene Aprons
(2)
Ansell Hycar Aprons
(2)
North Silver Shield Apron
(1)
Ansell Vinyl Aprons
(1)
Polyethylene-Coated Polypropylene Apron
(1)
Dupont Tyvek Apron
(1)
Disposable Polyethylene Aprons
(1)
PVC/Nylon Aprons
(1)
Brand:
Ansell
(10)
Cryo-Apron
(3)
Endurosaf
(3)
[+]
Show more
(3)
Aluminized Zetex
(3)
Tyvek
(1)
North Silver Shield
(1)
[-]
Show less
Product Type:
Aprons
(22)
Price Range:
$0 to $100
(18)
$100 to $299
(6)
Availability:
In Stock
(23)
Color:
Blue
(10)
White
(4)
Silver
(4)
[+]
Show more
(2)
Yellow
(3)
Black
(3)
[-]
Show less
Material:
Vinyl
(6)
Nylon
(3)
Polyethylene
(3)
[+]
Show more
(6)
Enduro 2000
(3)
Aluminized Zetec
(3)
Neoprene
(2)
Hycar
(2)
PVC/Nylon
(1)
Norfoil
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Safety Apparel
> Aprons
Aprons
Choose from our comprehensive line of Aprons and other Apparel products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Cryo-Apron aprons, 24"W x 42"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-38
In stock.
$156.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron Aprons
PVC/Nylon Aprons, 50"L x 35"W, 12/case
Product Type Aprons,Description Apron,Color Yellow
EW-81501-11
In stock.
$105.00 USD/CASE of 12
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
PVC/Nylon Aprons
Vinyl Coat Apron; Thickness, 8-mil; Size, Large
Product Type Aprons,Description Apron,Color Blue
EW-81620-56
In stock.
$24.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Vinyl Coat Apron
Cryo-Apron aprons, 24"W x 36"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-36
In stock.
$145.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron
Aprons
Polyethylene-Coated Polypropylene Aprons, 24" x 42"
Product Type Aprons,Description Apron,Color White
EW-86227-50
In stock.
$70.00 USD/CASE of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Polyethylene-Coated Polypropylene Apron
Ansell Endurosaf
Aprons, Blue
Product Type Aprons,Description Apron,Color Blue
EW-81620-05
In stock.
$19.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Endurosaf Aprons
Ansell Vinyl Apron, 6-mil, 35" x 45", 12/pk
Product Type Aprons,Description Apron,Color Blue
EW-81620-60
In stock.
$28.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Vinyl Aprons
Ansell Neoprene Apron, 35" x 45", black
Color Black,Material Neoprene,Brand Ansell
EW-86479-01
In stock.
$15.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Neoprene Aprons
Cryo-Apron aprons, 24"W x 48"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-40
In stock.
$187.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron Aprons
Tyvek apron, 36"L x 28"W, 100/cs
Product Type Aprons,Description Apron,Color White
EW-86225-80
In stock.
$133.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Dupont
Tyvek
Apron
Vinyl Coat Apron; Thickness, 8-mil; Size, Medium
Product Type Aprons,Description Apron,Color Blue
EW-81620-54
In stock.
$24.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Vinyl Coat Apron
Aluminized Zetex apron, 36"L x 24"W
Product Type Aprons,Description Apron,Color Silver
EW-86364-50
In stock.
$71.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Aluminized Zetex
Apron
Ansell Hycar Apron 35"x45" Heavy Weight 18 Oz
Product Type Aprons,Description Heavy Weight,Color Black
EW-86502-05
In stock.
$19.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Hycar Aprons
Vinyl Coat Apron; Thickness, 6-mil; Size, Large
Product Type Aprons,Description Apron,Color Blue
EW-81620-50
Usually ships in 6 days.
$14.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Vinyl Coat Apron
Disposable Polyethylene Aprons, 28"W X 46"L, 100/bag
Product Type Aprons,Description Disposable,Color White
EW-81620-23
In stock.
$9.50 USD/BAG
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Disposable Polyethylene Aprons
24
results (showing
1
-
15
)
1
2
Next >
>>
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Safety Apparel
> Aprons
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title