United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Titrators
>
Karl Fischer
> Aquamax Coulometric Karl Fischer Titrator
Aquamax Coulometric Karl Fischer Titrator
Use in the laboratory or out in the field!
Greaseless glassware provides quick and effortless assembly and disassembly
Built-in high-speed printer provides hard copy results and duplicates if required
The Cou-Lo Aquamax titrator sets new standards in versatility and ease of operation for determining water content in liquids, gases, and powders. The patent-pending ACE (Automatic Compensation of Errors) control system guarantees that the actual electrolysis current produced and the count rate displayed are always correctly synchronized, regardless of changes to the electrolysis cell resistance. This system provides the correct result even if the generator is partially blocked, minimizes errors in daily calibration, and increases the number of samples you can analyze. Use for ASTM D1533, D4928, D6304, IP 386, API MPMS Chapter 10.9, and BS 60814 test methods.
A truly versatile instrument, the Cou-Lo Aquamax displays results in %, ppm, mg/kg, and µg of water. Delayed start, automatic three-fold end point indication, and four different calculation modes accommodate a variety of applications. Printed results show statistics, data input parameters, sample ID numbers, and time-and-date stamp. Results can also be downloaded via the optional Results Manager software package into a PC spreadsheet format.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Aquamax Coulometric Karl Fischer Titrator
Product Type Karl Fischer Titrators,Display LCD,Power VAC 90 to 264; 12 VDC car adapter
EW-94101-00
Call 1-888-358-4705 for availability.
$6,500.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Aquamax Coulometric Karl Fischer Titrator
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Titrators
>
Karl Fischer
> Aquamax Coulometric Karl Fischer Titrator
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title