log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Lab Equipment > Titrators > Volumetric and Potentiometric > Aquamax Total Acid Number (TAN) Titrator
Aquamax Total Acid Number (TAN) Titrator
Aquamax Total Acid Number (TAN) Titrator
  • Conforms to ASTM D664 for determination of acidic constituents in petroleum products, lubricants and transformer insulating oils
  • Three pre-programmed methods for TAN content simplifies setup
  • Datalogger automatically stores up to 55 analysis results for easy recall
  • High precision syringe provides superior dispensing resolution
The Aquamax TAN Titrator is designed for the determination of acidic constituents in petroleum products, lubricants, and transformer insulating oils and conforms to ASTM D664. Titrator is easy-to-operate. Simply calibrate, run a blank, select the correct pre-programmed TAN analysis, and the titrator performs the analysis. Intuitive function guide display walks you through operation. Results are displayed on screen in units of KOH/g and are also stored via internal datalogger.
Flexible data management options allow for export of readings to a PC or printer. TiCOM software (order separately) allows viewing and printing of sample input, blank, and titration measurement data. Reports on data can be generated directly and exported to Excel or Access. Optional thermal printer accessory allows for the printing of results data. Order separately.
 Product Detail
Sort
 Availability
 Price
EW-94100-35 Aquamax Total Acid Number (TAN) Titrator
Aquamax Total Acid Number (TAN) Titrator
Product Type Acid-Base/Alkalinity Titrators,Display LCD,Power VAC 90 to 164
EW-94100-35
Usually ships in 7 days.
$6,150.25 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Lab Equipment > Titrators > Volumetric and Potentiometric > Aquamax Total Acid Number (TAN) Titrator
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title