Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Titrators
>
Karl Fischer
> Aquamax Volumetric Karl Fischer Titrator
Aquamax Volumetric Karl Fischer Titrator
Versatile instrument for the volumetric determination of water content using the KF method
Very easy to use - function guide display walks you through operation
Novel syringe control eliminates the common problem of bubble formation in the reagent circuit
User-friendly interface and intuitive keypad make operation of the Aquamax Volumetric KF Titrator simple to grasp. Titrator features two high-accuracy 5-mL syringes with 40,000 step resolution for superior dosage accuracy. One syringe can be used for the KF reagent and the other can be used for dosing of standard titre, different factor reagents, or specialty reagents. Flexible titrator accepts several different reagent brands.
Titrator is programmable and stores up to 10 different titration and standardization programs. Automatically monitor and compensate for drift during titration - pop up annunciator will prompt the user if drift exceeds programmed limit values. Built-in datalogging capability stores up to 55 analysis results. Optional communications software or printer allow for the viewing or printing of stored data for data management flexibility.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Aquamax Volumetric Karl Fischer Titrator
Product Type Volumetric Titrators,CE Compliance Yes,Brand Aquamax
EW-94100-01
Usually ships in 7 days.
$6,150.25 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Aquamax Volumetric Karl Fischer Titrator
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Titrators
>
Karl Fischer
> Aquamax Volumetric Karl Fischer Titrator
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity