log in or register
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Heating Equipment > Sterilizers > Sterilizer Accessories > Autoclave Deodorizer Capsules
Autoclave Deodorizer Capsules
Autoclave Deodorizer Capsules
  • Absorb and remove autoclave odors rather than just covering them
  • Choose apple or lemon scented capsules
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture dissolves the capsule and releases a clean, fresh scent.
 Product Detail
Sort
 Availability
 Price
Representative photo only Autoclave Deodorizer Lemon Scented 240 Capsules
Autoclave Deodorizer, Lemon Scented 240 Capsules
Product Type Sterilizer Accessories
EW-44468-28
In stock.
$230.00 USD/CASE
Qty:
Representative photo only Autoclave Deodorizer Apple Scented 240 Capsules
Autoclave Deodorizer, Apple Scented 240 Capsules
Product Type Sterilizer Accessories
EW-44468-27
Usually ships in 7 days.
$230.00 USD/CASE
Qty:
2 results (showing 1 - 2 )
Per Page 
Home  >  Heating Equipment > Sterilizers > Sterilizer Accessories > Autoclave Deodorizer Capsules
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title