United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(1)
$300 to $499
(1)
$500 to $799
(1)
[+]
Show more
(1)
$2,000 and Over
(1)
[-]
Show less
Temp sensor:
platinum RTD
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Melting Point
>
Digital Melting Point Apparatuses
> Automatic Digital Melting Point Apparatus
Automatic Digital Melting Point Apparatus
High-resolution camera captures the melting point-no operator needs to be present
Captures a video image of the melting process with the use of a PC
Conforms to US Pharmacopeia (USP) and GLP requirements
This digital unit automatically determines the melting point of chemical substances. A built-in camera continuously captures real-time images of the samples; digital image processing determines when melting occurs. Intuitive setup menu and front-panel touch screen make operation quick and easy; simply select the start temperature, remp reate, stop temperature, and press start. PID-controlled temperature ramping provides an extremely linear increase in temperature. Audible and visual alarms indicate temperature stabilization, end of melt, and temperature outside of low/high limits that are user selected. Analyze up to three samples simultaneously.
View graphical data of the melting event on the front-panel display, or display real-time, high-resolution images of your samples by connecting the unit to a PC through the USB connection. Included MeltView software allows you to play back a melting event as a continuous movie or view it frame-by-frame for detailed analysis. Print out a hard copy of text and graphs using the RS-232 port and the optional printer (sold seperately). Printouts include technician identification and space for a signature.
For samples that melt in a unique way, such as samples that explode, boil away, or bubble up the tube, a wide observation window with a illuminated magnification lens allows manual observation of melting point.
Additional features include three user-configurable timers that can be used when the instrument is not needed for melting point determinations, and an external RTD input (2-, 3-, or 4-wire 100ΩPt) to use the unit as a thermometer when not in use for melting point.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Automatic Digital Melting Point Apparatus
Temp sensor platinum RTD,Display 5.8" color LCD touch-screen,Power VAC 90 to 264
EW-03012-90
Usually ships in 4 days.
$4,000.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Automatic Digital Melting Point Apparatus
Replacement capillaries, 300/pack
EW-03012-92
Usually ships in 4 days.
$225.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Automatic Digital Melting Point Apparatus
Melting Point Standards
EW-03012-93
Usually ships in 4 days.
$435.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Automatic Digital Melting Point Apparatus
Optional Printer with cable and paper
EW-03012-91
Usually ships in 4 days.
$625.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Automatic Digital Melting Point Apparatus
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Melting Point
>
Digital Melting Point Apparatuses
> Automatic Digital Melting Point Apparatus
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title