log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Gas Detectors > Bacharach GDC-350 Fixed-Mount Digital Gas Monitors
Bacharach GDC-350 Fixed-Mount Digital Gas Monitors
Bacharach GDC-350 Fixed-Mount Digital Gas Monitors
  • Prewired power supply with cord set for out-of-box operation
Start monitoring quickly and easily—prewired power cord and easy mounting provides out-of-box operation. Detect one or two gases in an area with these fixed-mount gas monitors. Rugged PVC enclosure makes these monitors suitable for a wide range of applications. Calibration Extending Firmware (CEF) enhanced sensors account for the aging of the CO, NO, and NO2 sensors, decreasing the frequency of calibration. Front-panel LEDs and audible alarms indicate alarm condition. Two alarm relays control external devices such as fans, or connect to building management systems for notification.
 Product Detail
Sort
 Availability
 Price
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Oxygen
Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-81
Call 1-888-358-4705 for availability.
$745.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Nitric oxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitric oxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-83
Call 1-888-358-4705 for availability.
$870.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Carbon monoxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-80
Call 1-888-358-4705 for availability.
$720.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Oxygen Sulfur dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen/Sulfur dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-92
Call 1-888-358-4705 for availability.
$1,085.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Nitrogen dioxide Sulfur dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitrogen dioxide/Sulfur dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-94
Call 1-888-358-4705 for availability.
$1,210.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Carbon monoxide oxygen
Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide/oxygen
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-85
Call 1-888-358-4705 for availability.
$885.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Carbon monoxide Sulfur dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide/Sulfur dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-88
Call 1-888-358-4705 for availability.
$1,060.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Oxygen Nitric oxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen/Nitric oxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-91
Call 1-888-358-4705 for availability.
$1,035.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Nitrogen dioxide Nitric oxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitrogen dioxide/Nitric oxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-93
Call 1-888-358-4705 for availability.
$1,160.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Sulfur dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Sulfur dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-84
Call 1-888-358-4705 for availability.
$920.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Carbon monoxide Nitric oxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide/Nitric oxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-87
Call 1-888-358-4705 for availability.
$1,010.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Nitric oxide Sulfur dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitric oxide/Sulfur dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-95
Call 1-888-358-4705 for availability.
$1,210.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Carbon monoxide Nitrogen dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide/Nitrogen dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-86
Call 1-888-358-4705 for availability.
$1,010.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Oxygen Nitrogen dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen/Nitrogen dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-89
Call 1-888-358-4705 for availability.
$1,035.00 USD/EACH
Qty:
Representative photo only Bacharach GDC 350 Fixed Mount Gas Monitor Nitrogen dioxide
Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitrogen dioxide
Product Type Fixed-Mount Gas Detection,Alarm LED indicators and audible,Output Two SPDT relays rated for 5 amps at 250 VAC
EW-81973-82
Call 1-888-358-4705 for availability.
$870.00 USD/EACH
Qty:
15 results (showing 1 - 15 )
Per Page 
Home  >  Safety Products > Gas Detectors > Bacharach GDC-350 Fixed-Mount Digital Gas Monitors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title