Home  >  Bath & Circulator Accessories
Bath & Circulator Accessories
Bath & Circulator Accessories
Controlling bath fluid temperature can be a challenge, but Cole-Parmer offers a variety of accessories for your bath or circulator product.
Bath fluid allows you to maintain a constant temperature within your bath, from -65°C to 280°C. Use the following guide to select the one that is right for your application:
  • Silicone 210 fluid allows you to maintain temperatures between -65 and 280°C and has specific gravity of 0.98
  • Dynalene® HC-50 fluid has a temperature range of -50 to 58°C and a specific gravity of 0.85
  • Silicone 200 20 (hot) bath fluid creates an environment capable of reaching temperatures between 10 and 149°C with a specific gravity of 0.95
  • Silicone 710 (hot) bath fluid allows you to maintain temperatures between 150 and 250°C with a specific gravity of 1.11
Hollow ball bath covers provide insulation for your bath and reduce your vapor loss considerably. Insulating balls are made of HDPE and form a tight cover over your fluid. These hollow bath balls can reduce heat exchange by more than 75 percent.
Our selection of holders and racks hold your sample in place and increase the flexibility of your bath, protecting the bath fluid.
 Product Detail
Sort
 Availability
 Price
Representative photo only Silicone 210 Bath Fluid 65 to 280 C 1 gal 3 8 L
Silicone 210 Bath Fluid, -65 to 280 C; 1 gal (3.8 L)
Product Type Bath oil,Min temperature º C -65,Max temperature º C 280
EW-01294-60
Call 1-888-358-4705 for availability.
$415.00 USD/GALLON
Qty:
EW-08796-00 Lab algicide, 8 oz bottle
Concentrated Lab Algicide/Algaecide, 8 oz (250 mL)
Product Type Algicide,Brand PolyScience
EW-08796-00
Usually ships in 11 days.
$59.00 USD/EACH
Qty:
Representative photo only Plexiglas tank 27 5 liter capacity measures 20 L x 12 W x 8 H
Plexiglas tank, 27.5-liter capacity; measures 20"L x 12"W x 8"H
Product Type Water Bath,Bath type Tank Bath,Capacity Liters 27.5
EW-01137-20
In stock.
$312.00 USD/EACH
Qty:
Representative photo only Silicone 200 20 Bath Fluid 10 to 149 C 1 gal 3 8 L
Silicone 200 20 Bath Fluid, 10 to 149 C; 1 gal (3.8 L)
Product Type Bath oil,Min temperature º C -10,Max temperature º C 149
EW-01294-40
In stock.
$565.00 USD/GALLON
Qty:
EW-12110-00 PolyScience T 13R, Refrigerated Tank, 13 L (3.8 gal.); 120VAC/60Hz
PolyScience T 13R, Refrigerated Tank, 13 L (3.4 gal.); 120VAC/60Hz
Product Type Circulator (Cooling/Heating),Capacity Liters 13,Min temperature º C 0
EW-12110-00
Call 1-888-358-4705 for availability.
$1,825.00 USD/EACH
Qty:
EW-06821-04 Polypropylene Bath Balls
EW-06821-04
Usually ships in 8 days.
$156.00 USD/PACKAGE
Qty:
Representative photo only Bath Fluid Ethylene Glycol 25 to 100 C case of five 1 2 gal
Bath Fluid, Ethylene Glycol, -25 to 100 C, case of five 1/2-gal
Product Type Bath oil,Min temperature º C - 25,Max temperature º C 100
EW-01294-31
In stock.
$109.00 USD/CASE
Qty:
EW-06821-02 Polypropylene Bath Balls
EW-06821-02
In stock.
$102.00 USD/PACKAGE
Qty:
EW-01137-16 Large-Volume Plexiglas<small><sup>&reg;</sup></small> tank; Capacity 16 Liters
Large-Volume Plexiglas tank; Capacity 16 Liters
Product Type Water Bath,Bath type Tank Bath,Capacity Liters 16
EW-01137-16
In stock.
$202.00 USD/EACH
Qty:
EW-06821-00 Polypropylene Bath Balls
EW-06821-00
In stock.
$235.00 USD/PACKAGE
Qty:
EW-06821-08 Polypropylene Bath Balls
EW-06821-08
Usually ships in 8 days.
$424.00 USD/PACKAGE
Qty:
EW-12149-50 Julabo SW Lift-up Transparent cover
Julabo SW Lift-up Transparent cover
Product Type Accessory,Max temperature º C 80 deg C
EW-12149-50
Usually ships in 6 days.
$280.00 USD/EACH
Qty:
EW-01137-00 Plexiglas<small><sup>&reg;</sup></small> tank, 8-liter capacity; measures 4-3/4"L x 17-7/4"W x 5-15/16"H
Plexiglas tank, 8-liter capacity; measures 4-3/4"L x 17-7/4"W x 5-15/16"H
Product Type Water Bath,Bath type Tank Bath,Capacity Liters 8
EW-01137-00
Call 1-888-358-4705 for availability.
$180.00 USD/EACH
Qty:
Representative photo only Plexiglas tank 6 liter capacity measures 4 3 4 L x 13 7 8 W x 5 15 16 H
Plexiglas tank, 6-liter capacity; measures 4-3/4"L x 13-7/8"W x 5-15/16"H
Product Type Water Bath,Bath type Tank Bath,Capacity Liters 6
EW-01137-02
In stock.
$159.00 USD/EACH
Qty:
Representative photo only Dynalene HC 50 Bath Fluid 50 to 58 C 1 gal 3 8 L
Dynalene HC-50 Bath Fluid, -50 to 58 C; 1 gal (3.8 L)
Product Type Bath oil,Min temperature º C -50,Max temperature º C 58
EW-01297-00
In stock.
$187.00 USD/GALLON
Qty:
31 results (showing 1 - 15 )
Per Page 
Home  >  Bath & Circulator Accessories
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title