Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
> Benchtop Analytical Mill
Benchtop Analytical Mill
Hands-free grinding of small samples
Grind pharmaceuticals, seeds, frozen tissue, pigment blends, and rocks
Two-position rocker switch allows either pulse or continuous operation
Rugged polycarbonate mill with 304 stainless steel blade rapidly grinds small samples using a 1/5-hp motor. Grinding chamber disassembles to allow complete sample recovery and cleaning. Resettable circuit breaker protects the motor if the sample jams the blade. A built-in switch prevents operation unless all parts are completely assembledlight on the rocker switch indicates when unit is ready for use.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Analytical Mill
Product Type Analytical Mills,Power VAC 110
EW-04305-00
Call 1-888-358-4705 for availability.
$1,280.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Benchtop Analytical Mill
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
> Benchtop Analytical Mill
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity