log in or register
Brand:
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Benchtop Ultraviolet Transilluminators
Benchtop Ultraviolet Transilluminators
  • Compact size saves space over larger high-performance models
  • UV-blocking cover is hinged and height adjustable to suit the user and provide easy access to the surface
  • UVG long-life filters are rated for 5000 hours of use
Midrange (302 nm) models are available in single- or dual-intensity configuration. Dual intensity High setting is ideal for analytical documentation. Low setting reduces photonicking or photobleaching of gel samples while doing preparative work. Dual- and triple-wavelength models cover a wide range of applications reducing the need for multiple units. Use the longwave setting for viewing stained gels, the midrange setting for sensitive nucleic acid visualization, and the shortwave setting to irradiate samples. Safety cover blocks 99% of harmful UV radiation.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 230VAC
EW-97623-18
Usually ships in 10 days.
$1,764.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt UV Transilluminator 302nm Wavelength with 15cm X 15cm Filter 230VAC
Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 15cm X 15cm Filter, 230VAC
Product Type Ultraviolet Transilluminators,Brand Cole-Parmer
EW-97623-06
Usually ships in 10 days.
$1,180.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 20cm x 20cm Filter 115VAC
EW-97623-08
Usually ships in 10 days.
$1,415.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 230VAC
Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter, 230VAC
Product Type Ultraviolet Transilluminators,Brand Cole-Parmer
EW-97623-22
Usually ships in 10 days.
$1,708.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Dual Wavelength UV Transilluminator 302nm and 365nm Wavelength with 20cm x 20cm Filter 230VAC
EW-97623-26
Usually ships in 15 days.
$1,764.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 115VAC
EW-97623-16
Usually ships in 10 days.
$1,764.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 20cm x 20cm Filter 230VAC
EW-97623-10
Usually ships in 10 days.
$1,415.00 USD/EACH
Qty:
Representative photo only Triple Wavelength 8 Watt UV Transilluminator 254nm 302nm 365nm Wavelength with 20cm x 20cm Filter 115VAC
EW-97623-32
Call 1-888-358-4705 for availability.
$2,338.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt UV Transilluminator 302nm Wavelength with 15cm x 15cm Filter 115VAC
Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 15cm x 15cm Filter, 115VAC
Product Type Ultraviolet Transilluminators,Brand Cole-Parmer
EW-97623-04
Call 1-888-358-4705 for availability.
$1,180.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Dual Wavelength UV Transilluminator 302nm and 365nm Wavelength with 20cm x 20cm Filter 115VAC
EW-97623-24
Call 1-888-358-4705 for availability.
$1,764.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Dual Wavelength UV Transilluminator 302nm and 365nm Wavelength with 21cm x 26cm Filter 230VAC
EW-97623-30
Usually ships in 10 days.
$2,057.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Dual Wavelength UV Transilluminator 302nm and 365nm Wavelength with 21cm x 26cm Filter 115VAC
EW-97623-28
Usually ships in 10 days.
$2,057.00 USD/EACH
Qty:
Representative photo only Triple Wavelength 8 Watt UV Transilluminator 254nm 302nm 365nm Wavelength with 21cm x 26cm Filter 230VAC
EW-97623-38
Usually ships in 10 days.
$2,477.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 115VAC
Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter, 115VAC
Product Type Ultraviolet Transilluminators,Brand Cole-Parmer
EW-97623-20
Usually ships in 10 days.
$1,708.00 USD/EACH
Qty:
Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 15cm x 15cm Filter 230VAC
EW-97623-02
Usually ships in 10 days.
$1,242.00 USD/EACH
Qty:
20 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title