Beta Press for Solid Samples
Beta Press for Solid Samples
This unique precision solid sampling device makes KBr pellets in a 316 stainless stell barrel with two hardened and highly polished 316 stainless steel bolts. Remove bolts and mount beta press directly onto the spectrophotometer for analysis.
 Product Detail
Sort
 Availability
 Price
Representative photo only Beta press Includes barrel bolts slide holder and instructions for use with Scanning Infrared Spectrophotometer
EW-83002-00
Usually ships in 12 days.
$105.50 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title