log in or register
Category:
Brand:
Price Range:
Availability:
Color:
Clear (4)
Material:
HDPE (9)
LDPE (1)
Capacity (gal.):
1-5 (3)
10-20 (4)
Capacity (oz):
24 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Laboratory Supplies & Consumables > Bags > Biohazard Bags
Biohazard Bags
Biohazard Bags
Biohazard bags may seem like a quick solution to a potentially messy problem but they are not without their responsibilities. By order of OSHA, biohazard bags must be either red, bright orange or clear. The bags must be clearly marked "biohazard waste," or with the universal biohazard waste symbol or both. Cole-Parmer offers the highest quality bags for all your application needs. We have biohazard bags as large as 50 gallons down to 1/2 gallon size. All styles include a sterilization indicator patch that darkens when autoclaved.
For sampling applications, Whirl-Pak sample bags are an industry standard. Use these sterile LDPE bags to safely hold your solids and liquids. The bags are easy to use, simply flip bag around wire/tape and bend wire to seal. Whirl-Pak sampling bags are available with or without a white labeling area; other options include bags with sponge for surface sampling, high-temperature bags, and thiosulfate tablet bags for water sampling.
For general purpose use, zip-lip bags are available in clear LDPE and in amber PE. Use amber sample bags for light-sensitive samples. Being responsible waste by using biohazard bags and other safety equipment is essential to worker safety.
 Product Detail
Sort
 Availability
 Price
EW-06494-00 Biohazard Bag 14" x 19", 200 per Case
Biohazard Bag 14" x 19", 200 per Case
Product Type Biohazard Bags,Material HDPE,Capacity gal. 4
EW-06494-00
In stock.
$119.00 USD/CASE
Qty:
Representative photo only Biohazard Bag 31 x 38 200 per Case
Biohazard Bag 31" x 38", 200 per Case
Product Type Biohazard Bags,Material HDPE,Capacity gal. 30
EW-06494-06
Call 1-888-358-4705 for availability.
$400.00 USD/CASE
Qty:
Representative photo only Biohazard Bag 25 x 35 200 per Case
Biohazard Bag 25" x 35", 200 per Case
Product Type Biohazard Bags,Material HDPE,Capacity gal. 12
EW-06494-04
In stock.
$300.00 USD/CASE
Qty:
EW-06496-50 Bag shown with wire stand 06496-60
Desktop Biohazard Bag
Product Type Biohazard Bags,Material HDPE,Capacity gal. 0.5
EW-06496-50
Usually ships in 13 days or sooner.
$20.00 USD/PACKAGE
Qty:
Representative photo only Heavy Duty Biohazard Disposal Bags 25 W x 35 L
Heavy-Duty Biohazard Disposal Bags, 25"W x 35"L
Product Type Biohazard Bags,Material Polypropylene,Capacity gal. 12
EW-06495-04
In stock.
$495.00 USD/PACKAGE
Qty:
Representative photo only Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch 24 x 30
Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch, 24" x 30"
Product Type Biohazard Bags,Material HDPE,Capacity gal. 12
EW-06497-40
Usually ships in 12 days.
$134.00 USD/PACKAGE of 100
Qty:
Representative photo only Heavy Duty Biohazard Disposal Bags 19 W x 23 L
Heavy-Duty Biohazard Disposal Bags, 19"W x 23"L
Product Type Biohazard Bags,Material Polypropylene,Capacity gal. 9
EW-06495-02
In stock.
$265.00 USD/PACKAGE
Qty:
Representative photo only Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch 10 x 15
Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch, 10" x 15"
Product Type Biohazard Bags,Material HDPE,Capacity gal. 1
EW-06497-30
In stock.
$53.00 USD/PACKAGE of 100
Qty:
EW-06498-08 Whirl-Pak Biohazard Bag 6"x9"
Whirl-Pak Biohazard Bag 6"x9"
Product Type Biohazard Bags,Material LDPE,Capacity oz 24
EW-06498-08
In stock.
$126.00 USD/BOX
Qty:
Representative photo only Biohazard Bag 19 x 23 200 per Case
Biohazard Bag 19" x 23", 200 per Case
Product Type Biohazard Bags,Material HDPE,Capacity gal. 9
EW-06494-02
In stock.
$170.00 USD/CASE
Qty:
EW-06495-00 Heavy-Duty Biohazard Disposal Bags, 14"W x 19"L
Heavy-Duty Biohazard Disposal Bags, 14"W x 19"L
Product Type Biohazard Bags,Material Polypropylene,Capacity gal. 4
EW-06495-00
Usually ships in 14 days.
$186.00 USD/PACKAGE
Qty:
Representative photo only Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch 24 x 36
Scienceware Autoclavable HDPE Biohazard Disposal Bags with Sterilization Patch, 24" x 36"
Product Type Biohazard Bags,Material HDPE,Capacity gal. 20
EW-06497-50
In stock.
$150.00 USD/PACKAGE of 100
Qty:
Representative photo only Biohazard Bag 38 x 48 100 per Case
Biohazard Bag 38" x 48", 100 per Case
Product Type Biohazard Bags,Material HDPE,Capacity gal. 50
EW-06494-08
Usually ships in 13 days.
$310.00 USD/CASE
Qty:
13 results (showing 1 - 13 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Bags > Biohazard Bags
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title