United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Air Sampling
> Biotest Ergotouch Airborne Particle Counters
Biotest Ergotouch Airborne Particle Counters
Displays particle count totals of multiple particle sizes at once
Ideal for use in cleanrooms, indoor air quality, filter testing, and contamination migration studies. These handheld particle counters use a laser diode light source to detect particles between 0.3 and 10 µm. Ergonomic design allows one-handed portability. Both models feature automatic flow control, USB interface for data download, rechargeable batteries, high-impact housing, internal HEPA filter, and audible alarms.
Compliance:
NIST-calibrated according to Federal Standard 209, ASTM F-50; also meets JIS standard for counting accuracy.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Biotest ErgoTouch Pro Airborne Particle Counter
Product Type Particle Counters,Output USB and RS-232,Flow rate 0.1 cfm
EW-18450-25
In stock.
$3,942.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Biotest Ergotouch Airborne Particle Counters
Biotest ErgoTouch Airborne Particle Counter
Product Type Particle Counters,Output USB,Flow rate 0.1 cfm
EW-18450-20
Usually ships in 9 days.
$2,875.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Biotest Ergotouch Airborne Particle Counters
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Air Sampling
> Biotest Ergotouch Airborne Particle Counters
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title