Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Lab Equipment > Mixers and Blenders > Blenders and Paddle Blenders

Blenders and Paddle Blenders

Blenders and Paddle Blenders

Lab blenders are powerful and durable for grinding, blending, chopping, and processing. There are labs blenders used with both wet and dry ingredients, and are available in various sizes with varying speeds. Most lab blenders are designed specifically with the daily requirements of a laboratory in mind. Consider the following when selecting a mixer – the size of the container, house power, power supply, how many RPMs and what torque you need, among others. Waring® offers a vast array of lab blenders from the tradition light-duty ones to the four-liter container which processes compounds with high liquid content. Paddle lab blenders are used to blend solids and semisolids samples by applying pressure to samples within sterile, disposable bags with spring paddle action. Homogenization is produced through a combination of crushing and stirring, bringing out seated organisms from tissues, organs, powders, creams, ointments, etc. Explosion-proof lab blenders are used for safe mixing of unstable substances and hazardous materials. If you need help selecting lab blenders speak with a Cole-Parmer Technical Expert today.
We offer a broad selection of mixers and mixer products to suit almost any application. Proper selection of a mixer requires knowledge of certain application variables:
1) Container capacity;
2) Liquid viscosity (Dilatant, Newtonian, Pseudoplastic, Thixotropic);
3) Torque requirements;
4) Horsepower (hp) requirements;
5) Rotational speed (rpm) and diameter of mixing propeller;
6) Duty cycle;
7) Power supply.

Choose from a variety of product offerings within 6 categories of mixer types:

Laboratory Mixers: available with permanent magnet, universal, or airdrive motors. Choose different mixer mounts for mounting on tanks or stands. Our variety of impellers and paddles lend versatility to
the mixer you select.

Industrial Mixers: available with electric- or air-driven motors for up to 1-hp. Fixed-or variable-speed and direct- or gear-driven options are available.

Speciality Mixers include:
Sanitary Mixers: for all applications requiring extra clean processes.

Bulk-Container Mixers: mount atop tote containers and mix liquids that are in long-or short-term storage.

Handheld Mixers: for mixing thin or heavy liquids in open or closed containers.

Static Mixers: installed inline and mix fluids as you pump them through a pipeline. The fluid moves through the mixer in an alternating clockwise/ counterclockwise motion to ensure a homogeneous product. They are virtually maintenance free and need no spare parts.

Accessories are available for all our mixers. Our Application Specialists can assist you in choosing the right impellers for optimum mixing.
 Product Detail
Sort
 Availability
 Price
Representative photo only Waring SS four liter blender 120 VAC 60 Hz Measures 26 H

Waring SS four-liter blender; 120 VAC, 60 Hz; Measures 26"H

Product Type 4-Liter Blenders,Power VAC 120,Color White
EW-04240-01
In stock.
$2,042.00 USD/EACH

Qty:

EW-04249-00 Replacement Blade Assembly for 1 Liter Waring Blender

Replacement Blade Assembly for 1 Liter Waring Blender

Product Type Accessory,Model 003548,Brand Waring Products
EW-04249-00
In stock.
$27.00 USD/EACH

Qty:

EW-04240-60 Cool base container for four-liters containers

Cool base container for four-liters containers

Product Type Accessory,Model 2610C,Brand Waring Products
EW-04240-60
Call 1-888-358-4705 for availability.
$2,288.00 USD/EACH

Qty:

EW-04245-60 Waring variable-speed blender, 1-liter, glass 120v

Waring variable-speed blender, 1-liter, glass 120v

Product Type Variable Speed Blenders,Power VAC 120,Color White
EW-04245-60
In stock.
$818.00 USD/EACH

Qty:

Representative photo only Container PP 500ml

Container PP 500ml

Product Type Accessory,Model CAC64,Brand Waring Products
EW-04243-25
Usually ships in 10 days.
$138.00 USD/EACH

Qty:

Representative photo only Paddle Blender 50 To 400 mL Variable Speed Window Door Removable Paddles

Paddle Blender; 50 To 400 mL, Variable Speed, Window Door, Removable Paddles

Product Type Paddle Blenders,Description Standard Click and Clean® paddle blender, window door,Power VAC 110/220
EW-36850-13
Usually ships in 8 days.
$5,120.00 USD/EACH

Qty:

Representative photo only Waring 2 Speed Blender 1L Stainless Steel Container 240 VAC Standard Motor

Waring 2-Speed Blender, 1L, Stainless Steel Container, 240 VAC -- Standard Motor

Product Type 2 Speed Blenders,Description Standard motor,Power VAC 240
EW-04241-11
In stock.
$645.00 USD/EACH

Qty:

Representative photo only Waring blender 2 speed 1 liter glass 120v

Waring blender 2-speed, 1-liter, glass, 120v

Product Type 2 Speed Blenders,Power VAC 120,Color White
EW-04245-40
In stock.
$524.00 USD/EACH

Qty:

EW-36850-02 Seward Stomacher 80 Lab Blenders, compact, 230VAC

Seward Stomacher 80 Lab Blenders, compact, 230VAC

Product Type Paddle Blenders,Description Stomacher 80 Biomaster,Power VAC 230
EW-36850-02
Call 1-888-358-4705 for availability.
$5,475.00 USD/EACH

Qty:

EW-36850-04 Seward Stomacher 400C Lab Blenders, programmable, 110VAC

Seward Stomacher 400C Lab Blenders, programmable, 110VAC

Product Type Paddle Blenders,Description Stomacher 400C,Power VAC 110
EW-36850-04
Usually ships in 5 days.
$5,958.00 USD/EACH

Qty:

EW-36850-16 Note, this starter kit includes a blender with a stainless steel door.  For transparent door, order model 36850-17.

PADDLE BLENDER STARTER KIT; 400 mL Fixed speed blender with stainless steel door

Product Type Paddle Blenders,Power VAC 110/220,Brand Interscience Laboratories Inc
EW-36850-16
Usually ships in 8 days.
$3,656.00 USD/EACH

Qty:

EW-04241-10 Waring 2-Speed Blender, 1L, Stainless Steel Container, 120 VAC -- Standard Motor

Waring 2-Speed Blender, 1L, Stainless Steel Container, 120 VAC -- Standard Motor

Product Type 2 Speed Blenders,Description Standard motor,Power VAC 120
EW-04241-10
In stock.
$438.00 USD/EACH

Qty:

EW-36850-09 MiniMix Pictured

Paddle Blender; 5 To 100 mL, Variable Speed, Window Door, Removable Paddles

Product Type Paddle Blenders,Description MiniMix Click and Clean® paddle blender, window door,Power VAC 110/220
EW-36850-09
Usually ships in 8 days.
$4,350.00 USD/EACH

Qty:

Representative photo only Container w lid Glass 30 250ml

Container w/lid Glass 30-250ml

Product Type Accessory,Model 8470,Brand Eberbach Corp
EW-04243-28
In stock.
$220.00 USD/EACH

Qty:

EW-04248-30 Mini containers; 50 to 250 mL capacity

Mini containers; 50 to 250 mL capacity

Product Type Accessory,Model MC-3,Brand Waring Products
EW-04248-30
In stock.
$282.00 USD/EACH

Qty:

89 results (showing 1 - 15 )
Per Page 
Home  >  Lab Equipment > Mixers and Blenders > Blenders and Paddle Blenders

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title