United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Fluorometers
>
Thermo Scientific Quantech Fluorometer
> Borosilicate Cuvettes for Fluorometer
Borosilicate Cuvettes for Fluorometer
Photomultiplier tube (PMT) and microprocessor control provide excellent detection sensitivity
Automatically sets gain (sensitivity) and rangeeliminates time-consuming manual dial adjustments
Store up to nine multi-point standard curves in memory
Linear regression analysis permits direct concentration readings
Operates in English, Spanish, French, and German
RS-232 provides computer and printer connections
Retainer clip made of "memory metal" lets you use a variety of tube and cuvette sizes without special adapters
Compact and efficient, these fluorometers provide excellent analytical performance. Quantitate DNA and PCR products. Detect DNA contamination in drugs. Perform vitamin assays for pharmaceutical procedures. Environmental applications include the quantitative analysis of herbicides and pesticides in water and monitoring chlorophyll in lakes and water treatment plants.
Reduced source lamp replacement alone will show you the absolute economy of these fluorometers. For example, you can purchase a wide-band fluorometer (83075-20 or -25) with minimal operating costs for less than the cost of xenon or deuterium source lamp replacement for a single year.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Borosilicate Cuvettes for Turner Quantech
Fluorometer, Pack of Twelve
Product Type Fluorometers,Capacity 5 mL,Model 331775
EW-83075-46
Usually ships in 14 days.
$85.50 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Borosilicate Cuvettes for Fluorometer
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Fluorometers
>
Thermo Scientific Quantech Fluorometer
> Borosilicate Cuvettes for Fluorometer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title