Home  >  Laboratory Supplies & Consumables > Labels and Label Printers > Brady TLS PC Link Label Printer
Brady TLS PC Link Label Printer
Brady TLS PC Link Label Printer
  • From liquid nitrogen to hot water baths, thermal transfer printers provide a variety of solutions for your laboratory
  • TLS PC Link easily connects to PCs with USB cable
  • Pre-cut labels are designed specifically for laboratory applications
The TLS PC Link automatically recognizes label sizes and printer settings, eliminating wasted time and materials. The thermal transfer printing won’t smear, and is designed to print up to 500 labels a day. Print quality is 203 dpi.
Label printer comes with LabelMark™ label design software that enables user to easily design, edit, and print your labels. Software specifically written for laboratory applications feature programs that make creating custom labels efficient and accurate. Import graphics, create templates, print barcode symbols, and import label lists using ASCII and ODBC data import capabilities.
TLS PC Link connects to any computer and uses Brady labeling software, or any other Windows™ application. Runs on battery pack or optional AC adapter. Seventy-five foot label ribbons allow for large size banner printing. TLS PC Link includes Label Mark™ labeling software, rechargeable battery pack, power supply/battery charger, R6210 ribbon, sample labels, Quick Start Guide, communication cable, and Windows® driver. Order laboratory labels, replacement AC adapter, battery pack, and cleaning kit separately.
What's included: IdentiLab software, rechargeable battery pack, power supply/battery charger, black ribbon, sample labels, Quick Start Guide, PC cable, and Windows driver.
 Product Detail
Sort
 Availability
 Price
EW-65542-00 TLS PC Link Printer
TLS PC Link Printer
Product Type PC Link Printers,Color Gray
EW-65542-00
Usually ships in 5 days.
$705.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Labels and Label Printers > Brady TLS PC Link Label Printer
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title