Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Availability:
Volume (mL):
5 (1)
50 (1)
25 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Lab Equipment > Dispensers > Bottletop Dispensers > Brand Dispensette III Bottletop Dispensers > BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting

  • SafetyPrime valve recirculates reagent during priming to reduce spurting and conserve reagent.
  • Safety discharge system reduces the risk of accidental dispensing.
  • Valve block rotates 360° so that the volume display and bottle label are always visible.
  • Floating piston design for smooth, one-handed operation
  • Reinforced discharge tube with a threaded safety cap
  • Polypropylene cylinder sleeve helps protect the user in case of accidental breakage.
  • Digital models feature Easy Calibration for quick adjustments in the lab—without having to retest.
  • Can be calibrated for ISO/GLP compliance
  • Fit directly onto 45-mm screw-neck bottles. Come with polypropylene adapters for other neck sizes. Order bottles separately).
Dispensette® Organic Dispensers Ideal for the dispensing of concentrated acids, fuming acids, peroxides, and difficult organic solvents—such as THF and methylene chloride*. Bright yellow markings let you easily identify these organic bottletop dispensers.
 Product Detail
Sort
 Availability
 Price
Representative photo only BrandTech Dispensette Organic Bottletop Dispenser Analog Volume Setting 1 0 to 10 0 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 1.0 to 10.0 mL

Product Type Dispenser (Bottletop),Volume mL 10,Autoclavable Yes
EW-07902-38
In stock.
$465.00 USD/EACH

Qty:

Representative photo only BrandTech Dispensette Organic Bottletop Dispenser Analog Volume Setting 2 5 to 25 0 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 2.5 to 25.0 mL

Product Type Dispenser (Bottletop),Volume mL 25,Autoclavable Yes
EW-07902-42
Usually ships in 8 days.
$650.00 USD/EACH

Qty:

Representative photo only BrandTech Dispensette Organic Bottletop Dispenser Analog Volume Setting 0 5 to 5 0 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 0.5 to 5.0 mL

Product Type Dispenser (Bottletop),Volume mL 5,Autoclavable Yes
EW-07902-32
Usually ships in 5 days.
$465.00 USD/EACH

Qty:

EW-07902-44 BrandTech Dispensette<small><sup>&reg;</sup></small> Organic Bottletop Dispenser, Analog Volume Setting, 5 to 50 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 5 to 50 mL

Product Type Dispenser (Bottletop),Volume mL 50,Autoclavable Yes
EW-07902-44
Call 1-888-358-4705 for availability.
$660.00 USD/EACH

Qty:

Representative photo only BrandTech Dispensette Organic Bottletop Dispenser Analog Volume Setting 10 TO 100 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 10 TO 100 mL

Product Type Dispenser (Bottletop),Volume mL 100,Autoclavable Yes
EW-07902-46
Call 1-888-358-4705 for availability.
$1,020.00 USD/EACH

Qty:

5 results (showing 1 - 5 )
Per Page 
Home  >  Lab Equipment > Dispensers > Bottletop Dispensers > Brand Dispensette III Bottletop Dispensers > BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title