United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Brix scale range (Minimum):
0
(4)
45
(1)
28
(1)
[+]
Show more
(1)
58
(1)
[-]
Show less
Brix scale range (Maximum):
32
(2)
90
(1)
10
(1)
[+]
Show more
(3)
62
(1)
82
(1)
18
(1)
[-]
Show less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Refractometers
>
Handheld Manual Refractometers
> Brix Refractometers
Brix Refractometers
Durable units provide quick and easy measurements
Use these economical refractometers to take quick and easy measurements. Refractometers feature adjustable focus and a zero adjustment screw for calibration. Each refractometer is calibrated and adjusted to read at 68°F (20°C). Models with automatic temperature compensation (ATC) are ideal for applications where the temperatures of your sample vary.
Refractometers are made of corrosion-resistant brass and have rubber-insulated grips. Include hard case, calibration screwdriver, and instructions.
Brix Refractometers
measure soluble solid concentration of sugars, salts, proteins, and more in foods and liquids - ideal for quality control and R&D applications. Use to measure percent Brix of juices, beverages, and other sugar-based liquids.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Low-cost Brix refractometer with ATC; 0 to 10% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 0,Brix scale range Maximum 10
EW-81150-02
In stock.
$126.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Low-cost Brix refractometer with ATC, 0 to 32% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 0,Brix scale range Maximum 32
EW-81150-08
In stock.
$126.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Low-cost Brix refractometer; 0 to 32% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 0,Brix scale range Maximum 32
EW-81150-06
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Sugar/honey refractometer, 58 to 90% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 58,Brix scale range Maximum 90
EW-81150-14
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Low-cost Brix refractometer, 28 to 62% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 28,Brix scale range Maximum 62
EW-81150-10
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Low-cost Brix refractometer, 45 to 82% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 45,Brix scale range Maximum 82
EW-81150-12
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
Low-cost Brix refractometer, 0 to 18% Brix
Product Type Refractometer (Analog),Brix scale range Minimum 0,Brix scale range Maximum 18
EW-81150-04
In stock.
$106.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Brix Refractometers
7
results (showing
1
-
7
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Refractometers
>
Handheld Manual Refractometers
> Brix Refractometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title