United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$0 to $100
(3)
$100 to $299
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Process Equipment & Instrumentation
>
Steam Handling
>
Spirax Sarco Steam "Y" Strainers
> Bronze "Y" stainers
Bronze "Y" stainers
In-line strainer for added protection against dirt and pipe scale
20 mesh screen
Install in a vertical or horizontal pipeline
Typical applications:
Helps remove pipeline debris and protects pipeline components such as valves, pumps, and heat transfer equipment in steam systems.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Bronze "Y" strainer, 3/4" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406
F (208
C)
EW-07600-88
Usually ships in 7 days.
$115.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Bronze "Y" stainers
Bronze "Y" strainer, 1" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406
F (208
C)
EW-07600-90
Usually ships in 7 days.
$151.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Bronze "Y" stainers
Bronze "Y" strainer, 1/2" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406
F (208
C)
EW-07600-86
Usually ships in 7 days.
$82.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Bronze "Y" stainers
Bronze "Y" strainer, 3/8" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406
F (208
C)
EW-07600-84
Usually ships in 7 days.
$61.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Bronze "Y" stainers
Bronze "Y" strainer, 1/4" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406
F (208
C)
EW-07600-82
Usually ships in 7 days.
$61.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Bronze "Y" stainers
5
results (showing
1
-
5
)
Per Page
5
10
15
25
50
Home
>
Process Equipment & Instrumentation
>
Steam Handling
>
Spirax Sarco Steam "Y" Strainers
> Bronze "Y" stainers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title