Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Availability:
Condenser type:
Glass type:
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Buchi Rotavapor RII Rotary Evaporators

Buchi Rotavapor RII Rotary Evaporators

  • Durability and reliability with simple operation
  • Small footprint to maximize your lab space
  • Choice of three glassware configurations for application versatility
  • Glassware available with protective safety coating to minimize breakage
Designed specifically to meet standard distillation needs in the educational laboratory, these economically priced models feature analog controls for both rotation and bath temperature. Set rotation speed from 20 to 280 rpm. Heating bath features corrosion-resistant coating and operates from 20 to 95°C; quick-release electrical connection makes it particularly easy to remove for fluid removal or addition. Manual lift allows the evaporator to be raised or lowered by hand, making operation quick and safe.

Choose from three condenser assemblies for application versatility. Use the diagonal condenser assembly for standard distillations and where lab space limits height. The vertical condenser is used for standard distillations but takes up less bench space than the diagonal unit. The cold-trap condenser is
ideal for applications without cold water circulation.

Select from systems with standard glassware or glassware with Plastic + Glas® coating, which protects you against broken glass and dangerous reagents due to broken glass. Safety coating resists most solvents and acids, and is available on all glassware except evaporating flask.

Note: Direct exposure of Plastic + Glas glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.
 Product Detail
Sort
 Availability
 Price
Representative photo only RII Diagonal Rotary Evaporator System 115V

RII Diagonal Rotary Evaporator System, 115V

Condenser type Diagonal Evaporators,Glass type Uncoated,CE Compliance Yes
EW-38605-00
Call 1-888-358-4705 for availability.
$4,440.00 USD/EACH

Qty:

Representative photo only RII Safety Coated Vertical Rotary Evaporator System 115V

RII Safety-Coated Vertical Rotary Evaporator System, 115V

Condenser type Vertical Evaporators,Glass type Plastic and Glas coated,CE Compliance Yes
EW-38605-40
Usually ships in 3 days.
$4,770.00 USD/EACH

Qty:

Representative photo only RII Safety Coated Cold Trap Rotary Evaporator System 115V

RII Safety-Coated Cold-Trap Rotary Evaporator System, 115V

Condenser type Cold-trap Evaporators,Glass type Plastic and Glas coated,CE Compliance Yes
EW-38605-50
Usually ships in 7 days.
$5,080.00 USD/EACH

Qty:

Representative photo only RII Diagonal Rotary Evaporator System 220V

RII Diagonal Rotary Evaporator System, 220V

Condenser type Diagonal Evaporators,Glass type Uncoated,CE Compliance Yes
EW-38605-05
In stock.
$4,440.00 USD/EACH

Qty:

4 results (showing 1 - 4 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title