Home  >  Burettes (Burets)
Burettes (Burets)
Burettes (Burets)
Precision becomes easier when using burettes (burets). Cole-Parmer has a comprehensive line of these precision instruments to help you accurately measure liquids in your laboratory. Our wide range of standard burettes include both acrylic and Class A and Class B glass that are individually calibrated, feature enameled graduations, different styles of stopcock plugs and are calibrated to meet several ASTM specifications. To save money, try one of our self-zeroing burettes. These don’t require funneling and hazardous, wasteful spills are eliminated. Select from our extensive offering of bottletop and electronic burettes. Electronic burettes offer exact precise liquid measurement and have digital displays for easy reading. Capacity ranges are 9.9mL to 50mL and accuracy is achieved with a tolerance of ±0.2%. Adapters are offered with the individual burettes to meet various bottle neck sizes. A full-range of bottles are offered to meet your individual laboratory requirements. Speak to one of our Technical Experts who can help you with additional product specifications and guide you through selecting burettes that meet your requirements.
 Product Detail
Sort
 Availability
 Price
Representative photo only Titrette Digital Bottletop Burette Dispenser 50 mL
Titrette Digital Bottletop Burette/Dispenser; 50 mL
Volume mL 50,Brand Titrette,Product Type Bottletop Dispenser
EW-07910-53
In stock.
$1,032.00 USD/EACH
Qty:
EW-07879-30 Automatic self-zeroing, self-supporting burette; capacity, 25mL; subdivision, 0.1mL; reservoir, 1000mL
Automatic Self-Zeroing and Supporting Burette, 25 mL; 1/Pk
Capacity mL 25,Product Type Self-Zeroing Burettes
EW-07879-30
In stock.
$266.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Nalgene Self Zeroing Burette 25 mL 1 Pk
Thermo Scientific Nalgene Self-Zeroing Burette, 25 mL; 1/Pk
Capacity mL 25,Product Type Self-Zeroing Burettes,Brand Thermo Scientific Nalgene
EW-06209-20
In stock.
$135.00 USD/EACH
Qty:
Representative photo only Digitrate Pro Digital Burette Bottletop Dispenser 50 mL
Digitrate Pro Digital Burette, Bottletop Dispenser; 50 mL
Capacity mL 50,Product Type Digital Burettes,Brand Digitrate
EW-25205-10
Usually ships in 5 days or sooner.
$906.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Nalgene Self Zeroing Burette 50 mL 1 Pk
Thermo Scientific Nalgene Self-Zeroing Burette, 50 mL; 1/Pk
Capacity mL 50,Product Type Self-Zeroing Burettes,Brand Thermo Scientific Nalgene
EW-06209-30
In stock.
$150.00 USD/EACH
Qty:
EW-07910-51 Titrette<small><sup>&reg;</sup></small> Bottletop Burette, 25mL
Titrette Digital Bottletop Burette/Dispenser; 25 mL
Volume mL 25,Brand Titrette,Product Type Bottletop Dispenser
EW-07910-51
In stock.
$1,032.00 USD/EACH
Qty:
Representative photo only Automatic Self Zeroing and Supporting Burette 10 mL 1 Pk
Automatic Self-Zeroing and Supporting Burette, 10 mL; 1/Pk
Capacity mL 10,Product Type Self-Zeroing Burettes
EW-07879-10
In stock.
$253.00 USD/EACH
Qty:
EW-25205-00 Digitrate Pro, 30mL Electric Burette
Digitrate Pro Digital Burette, Bottletop Dispenser; 30 mL
Capacity mL 30,Product Type Digital Burettes,Brand Digitrate
EW-25205-00
Usually ships in 8 days or sooner.
$906.00 USD/EACH
Qty:
Representative photo only Automatic Self Zeroing and Supporting Burette 50 mL 1 Pk
Automatic Self-Zeroing and Supporting Burette, 50 mL; 1/Pk
Capacity mL 50,Product Type Self-Zeroing Burettes
EW-07879-40
In stock.
$307.00 USD/EACH
Qty:
Representative photo only Digitrate Electronic Burettes 50 mL capacity
Digitrate Electronic Burettes; 50 mL capacity
Capacity mL 50,Product Type Electronic Burettes,Brand Digitrate
EW-25201-15
In stock.
$955.00 USD/EACH
Qty:
Representative photo only Kimax Serialized and Certified Class A Glass Burette 50 mL 1 Pk
Kimax Serialized and Certified Class A Glass Burette, 50 mL; 1/Pk
Capacity mL 50,Product Type Class A Burettes,Brand Kimax
EW-34541-04
In stock.
$244.00 USD/EACH
Qty:
EW-34543-23 PYREX<small><sup>&reg;</sup></small> Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL; tolerance, &plusmn;0.05mL
Pyrex 2103 Class A Glass Burette, 50 mL; 1/Cs
Capacity mL 50,Product Type Class A Burettes,Brand Pyrex
EW-34543-23
In stock.
$164.00 USD/CASE of 1
Qty:
Representative photo only Cole Parmer Class B Burette w PTFE stopcock 50ML 1 ea
Cole-Parmer Class B Burette w/PTFE stopcock; 50ML, 1/ea
Capacity mL 50,Product Type Class B Burettes,Brand Cole-Parmer
EW-63100-38
In stock.
$80.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Nalgene Self Zeroing Burette 10 mL 1 Pk
Thermo Scientific Nalgene Self-Zeroing Burette, 10 mL; 1/Pk
Capacity mL 10,Product Type Self-Zeroing Burettes,Brand Thermo Scientific Nalgene
EW-06209-10
In stock.
$124.00 USD/EACH
Qty:
Representative photo only Cole Parmer Class B Burette w PTFE stopcock 25ML 1 ea
Cole-Parmer Class B Burette w/PTFE stopcock; 25ML, 1/ea
Capacity mL 25,Product Type Class B Burettes,Brand Cole-Parmer
EW-63100-36
Usually ships in 7 days or sooner.
$73.00 USD/EACH
Qty:
68 results (showing 1 - 15 )
Per Page 
Home  >  Burettes (Burets)