Burkert Rocker Valves
Burkert Rocker Valves
Choose the rocker or flipper two-, or three-way solenoid valve with different connection options to better suit your application needs. Rocker and flipper design are available in two–way direct-acting, NC; and three–way direct-acting, universal functions with any flow direction. The valve design features an isolation diaphragm that separates liquid from the actuator. This unique quality promotes heat transfer elimination as coil is not directly in contact with diaphragm. Both rocker and flipper valves have been proven in thousands of applications throughout the world and offer the highest life cycle available, over 100 million actuation cycles. Valves have minimal internal volume that is easily purged.
Use solenoid valves for chemical and laboratory applications. All valves are ideal for manifold mounting. Order manifolds at right.Rocker Valves have a miniature rocker that operates the isolated diaphragm to separate actuator from the media. Valve is only 16 mm wide. Each valve is equipped with an LED display and push-on tube connections. Valves include two 19" electrical leads.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Two Way Micro Solenoid Rocker Valve 1 4 28 UNF
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Product Type Solenoid Electric Valves (actuated),Cv 0.046,Max temperature º F 131
EW-98622-00
In stock.
$216.00 USD/EACH
Qty:
Representative photo only Cole Parmer Three Way Micro Solenoid Rocker Valve 1 4 28 UNF
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Product Type Solenoid Electric Valves (actuated),Cv 0.029,Max temperature º F 131
EW-98622-10
Call 1-888-358-4705 for availability.
$219.00 USD/EACH
Qty:
Representative photo only Cole Parmer Three Way Micro Solenoid Rocker Valve Manifold Mount
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, Manifold Mount
Product Type Solenoid Electric Valves (actuated),Cv 0.038,Max temperature º F 131
EW-98622-16
Call 1-888-358-4705 for availability.
$174.00 USD/EACH
Qty:
Representative photo only Cole Parmer Two Way Micro Solenoid Rocker Valve 1 8 NPT F
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 1/8 NPT(F)
Product Type Solenoid Electric Valves (actuated),Cv 0.071,Max temperature º F 131
EW-98622-02
Usually ships in 5 days.
$204.00 USD/EACH
Qty:
Representative photo only Cole Parmer Three Way Micro Solenoid Rocker Valve 3 32 Hose Barb
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 3/32" Hose Barb
Product Type Solenoid Electric Valves (actuated),Cv 0.29,Max temperature º F 131
EW-98622-14
Usually ships in 8 days.
$207.00 USD/EACH
Qty:
Representative photo only Cole Parmer Two Way Micro Solenoid Rocker Valve 3 32 Hosebarb
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 3/32" Hosebarb
Product Type Solenoid Electric Valves (actuated),Cv 0.046,Max temperature º F 131
EW-98622-04
Usually ships in 12 days.
$204.00 USD/EACH
Qty:
EW-98622-12 Cole-Parmer<small><sup>&reg;</sup></small> Three-Way Micro Solenoid Rocker Valve, 1/8 NPT(F)
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/8 NPT(F)
Product Type Solenoid Electric Valves (actuated),Cv 0.055,Max temperature º F 131
EW-98622-12
Call 1-888-358-4705 for availability.
$207.00 USD/EACH
Qty:
Representative photo only Cole Parmer Two Way Micro Solenoid Rocker Valve Manifold Mount
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, Manifold Mount
Product Type Solenoid Electric Valves (actuated),Cv 0.046,Max temperature º F 131
EW-98622-06
In stock.
$186.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title