United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Capacity (Liters):
0 to 1
(1)
Capacity (mL):
0 - 20
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Extraction Apparatus
>
Gerber-Method Fat Analysis System
> Butyrometers for Gerber-Method Fat Analysis
Butyrometers for Gerber-Method Fat Analysis
Get test results in 15 minutessaves you time and money
Use these butyrometers to measure the percent fat in dairy products. Butyrometers are precision calibrated and feature permanent graduations. Made of resistant borosilicate glass.
Order glass cups for Roeder and Van-Gulik method testing. Stoppers with special acid resistant rubber are available for all butyrometers. Both stopper 17425-15 and stopper pusher 17425-16 are required for milk and Kohler method testing; stoppers 17425-14 and -17 are required for Roeder and Van-Gulik method testing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Gerber-method Butyrometer; milk test
Product Type Extraction bottles,Material Glass,Capacity Liters 0.011
EW-17425-10
In stock.
$26.25 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Butyrometers for Gerber-Method Fat Analysis
Gerber-method Butyrometer; cheese test (Van-Gulik)
Product Type Extraction bottles,Material Glass
EW-17425-13
In stock.
$35.50 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Butyrometers for Gerber-Method Fat Analysis
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Extraction Apparatus
>
Gerber-Method Fat Analysis System
> Butyrometers for Gerber-Method Fat Analysis
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title